Skip to Content
Merck
All Photos(1)

Documents

637564

Sigma-Aldrich

N-Boc-3-pyrrolidinone

97%

Synonym(s):

N-(tert-Butoxycarbonyl)-3-pyrrolidinone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H15NO3
CAS Number:
Molecular Weight:
185.22
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

mp

34-38 °C (lit.)

storage temp.

−20°C

SMILES string

CC(C)(C)OC(=O)N1CCC(=O)C1

InChI

1S/C9H15NO3/c1-9(2,3)13-8(12)10-5-4-7(11)6-10/h4-6H2,1-3H3

InChI key

JSOMVCDXPUXKIC-UHFFFAOYSA-N

Application

N-Boc-3-pyrrolidinone can be used as starting material in the synthesis of spirocyclic tetrahydrofuran.
Used in a study of asymmetric hydrogen-transfer bioreduction of ketones with Leifsonia alcohol dehydrogenase.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

"Synthesis of a family of spirocyclic scaffolds: building blocks for the exploration of chemical space"
Kumar S, et al.
The Journal of Organic Chemistry, 78'(13), 6529-6539 (2013)
Tetrahedron Asymmetry, 16, 2539-2539 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service