Skip to Content
Merck
All Photos(3)

Key Documents

634786

Sigma-Aldrich

(S)-(+)-N-Boc-3-pyrrolidinol

97%

Synonym(s):

(S)-N-(tert-Butoxycarbonyl)-(+)-3-pyrrolidinol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H17NO3
CAS Number:
Molecular Weight:
187.24
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

optical activity

[α]20/D +26°, c = 1% in methanol

mp

60-64 °C (lit.)

functional group

hydroxyl

SMILES string

CC(C)(C)OC(=O)N1CC[C@H](O)C1

InChI

1S/C9H17NO3/c1-9(2,3)13-8(12)10-5-4-7(11)6-10/h7,11H,4-6H2,1-3H3/t7-/m0/s1

InChI key

APCBTRDHCDOPNY-ZETCQYMHSA-N

Application

(S)-(+)-N-Boc-3-pyrrolidinol can be used as a reactant to synthesize:
  • tert-Butyl 3-(2-bromophenoxy)pyrrolidine-1-carboxylate, which is employed as a key intermediate in the preparation of IkB-kinase IKK2 inhibitor.
  • Dimethoxy-pyrrolidylquinazoline , and peptidomimetic quinoline derivatives.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas A Chappie et al.
Journal of medicinal chemistry, 50(2), 182-185 (2007-01-19)
A papaverine based pharmacophore model for PDE10A inhibition was generated via SBDD and used to design a library of 4-amino-6,7-dimethoxyquinazolines. From this library emerged an aryl ether pyrrolidyl 6,7-dimethoxyquinazoline series that became the focal point for additional modeling, X-ray, and
Discovery of selective irreversible inhibitors for Bruton?s tyrosine kinase
P Zhengying, et al.
ChemMedChem, 2(1), 58-61 (2007)
Erik A A Wallén et al.
Journal of medicinal chemistry, 46(21), 4543-4551 (2003-10-03)
Isophthalic acid bis(l-prolyl-pyrrolidine) amide is a very potent prolyl oligopeptidase inhibitor, but it has a log P value of -0.2, which is very low for a compound targeted to the brain. Therefore, these types of compounds were further modified to
Exploiting the Differential Reactivities of Halogen Atoms: Development of a Scalable Route to IKK2 Inhibitor AZD3264
Murugan A, et al.
Organic Process Research & Development, 18(5), 646-651 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service