Skip to Content
Merck
All Photos(1)

Documents

609323

Sigma-Aldrich

Nitric-15N acid solution

~10 N in H2O, 98 atom % 15N

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H15NO3
CAS Number:
Molecular Weight:
64.01
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

98 atom % 15N

Quality Level

concentration

~10 N in H2O

bp

120.5 °C

density

1.422 g/mL at 25 °C

mass shift

M+1

storage temp.

−20°C

SMILES string

O[15N+]([O-])=O

InChI

1S/HNO3/c2-1(3)4/h(H,2,3,4)/i1+1

InChI key

GRYLNZFGIOXLOG-OUBTZVSYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Flame over circleCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Ox. Liq. 3 - Skin Corr. 1A

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service