Skip to Content
Merck
All Photos(1)

Key Documents

512443

Sigma-Aldrich

3-Chloro-L-tyrosine

97%

Synonym(s):

(2S)-2-Amino-3-(3-chloro-4-hydroxyphenyl)propanoic acid, Monochlorotyrosine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
4-(HO)C6H3(Cl)CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
215.63
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

optical activity

[α]24/D −2°, c = 1 in 1 M HCl

reaction suitability

reaction type: solution phase peptide synthesis

mp

249 °C (lit.)

application(s)

peptide synthesis

SMILES string

N[C@@H](Cc1ccc(O)c(Cl)c1)C(O)=O

InChI

1S/C9H10ClNO3/c10-6-3-5(1-2-8(6)12)4-7(11)9(13)14/h1-3,7,12H,4,11H2,(H,13,14)/t7-/m0/s1

InChI key

ACWBBAGYTKWBCD-ZETCQYMHSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laetitia Mouls et al.
Proteomics, 9(6), 1617-1631 (2009-03-03)
Protein oxidation is thought to contribute to a number of inflammatory diseases, hence the development of sensitive and specific analytical techniques to detect oxidative PTMs (oxPTMs) in biological samples is highly desirable. Precursor ion scanning for fragment ions of oxidized
Eline Thomson et al.
Free radical biology & medicine, 49(9), 1354-1360 (2010-07-22)
We aimed to determine whether myeloperoxidase (MPO) is the main peroxidase present in the airways of children with cystic fibrosis (CF) and to assess which oxidants it produces and whether they are associated with clinical features of CF. Children with
Michael Holzer et al.
Antioxidants & redox signaling, 14(12), 2337-2346 (2011-01-18)
Carbamylation of proteins through reactive cyanate has been demonstrated to predict an increased cardiovascular risk. Cyanate is formed in vivo by breakdown of urea and at sites of inflammation by the phagocyte protein myeloperoxidase. Because myeloperoxidase (MPO) associates with high-density
Mariusz G Fleszar et al.
Molecules (Basel, Switzerland), 25(21) (2020-11-11)
Quantification with satisfactory specificity and sensitivity of free 3-Nitro-l-tyrosine (3-NT), 3-Chloro-l-tyrosine (3-CT), and 3-Bromo-l-tyrosine (3-BT) in biological samples as potential inflammation, oxidative stress, and cancer biomarkers is analytically challenging. We aimed at developing a liquid chromatography-tandem mass spectrometry (LC-MS/MS)-based method
Matthew P Curtis et al.
Chemical research in toxicology, 24(3), 418-428 (2011-02-16)
The persistent activation of innate immune cells in chronic inflammation is gaining recognition as a contributing factor in a number of human diseases. A distinguishing feature of activated leukocytes at sites of inflammation is their production of reactive species such

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service