Skip to Content
Merck
All Photos(1)

Key Documents

510483

Sigma-Aldrich

Bis(ethylcyclopentadienyl)nickel(II)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ni(C5H4C2H5)2
CAS Number:
Molecular Weight:
244.99
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

liquid

reaction suitability

core: nickel

bp

90 °C (lit.)

density

1.137 g/mL at 25 °C (lit.)

SMILES string

[Ni].CC[C]1[CH][CH][CH][CH]1.CC[C]2[CH][CH][CH][CH]2

InChI

1S/2C7H9.Ni/c2*1-2-7-5-3-4-6-7;/h2*3-6H,2H2,1H3;

InChI key

VWHXTCKWIVCDGV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Atomic number of base material: 28 Nickel

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

228.2 °F - closed cup

Flash Point(C)

109 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Thin film photovoltaic devices have become increasingly important in efficiently harnessing solar energy to meet consumer demand.

Spintronics offer breakthroughs over conventional memory/logic devices with lower power, leakage, saturation, and complexity.

Spintronics offer breakthroughs over conventional memory/logic devices with lower power, leakage, saturation, and complexity.

Spintronics offer breakthroughs over conventional memory/logic devices with lower power, leakage, saturation, and complexity.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service