Skip to Content
Merck
All Photos(1)

Documents

488887

Sigma-Aldrich

Dioctyl phthalate-3,4,5,6-d4

98 atom % D

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6D4[CO2CH2(CH2)6CH3]2
CAS Number:
Molecular Weight:
394.58
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

98 atom % D

Quality Level

Assay

99% (CP)

refractive index

n20/D 1.486 (lit.)

bp

384 °C (lit.)

mp

−50 °C (lit.)

density

0.991 g/mL at 25 °C

mass shift

M+4

SMILES string

[2H]c1c([2H])c([2H])c(C(=O)OCCCCCCCC)c(c1[2H])C(=O)OCCCCCCCC

InChI

1S/C24H38O4/c1-3-5-7-9-11-15-19-27-23(25)21-17-13-14-18-22(21)24(26)28-20-16-12-10-8-6-4-2/h13-14,17-18H,3-12,15-16,19-20H2,1-2H3/i13D,14D,17D,18D

InChI key

MQIUGAXCHLFZKX-AMEAAFLOSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

228.2 °F - closed cup

Flash Point(C)

109 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaowei Liu et al.
Environmental science and pollution research international, 22(14), 10690-10698 (2015-03-11)
The first nationwide survey of six phthalates (diethyl phthalate (DEP); dimethyl phthalate (DMP); di-n-butyl phthalate (DBP); butyl benzyl phthalate (BBP); bis(2-ethylhexyl) phthalate (DEHP); din-octyl phthalate (DnOP)) in drinking waters from waterworks was conducted across seven geographical zones in China. Of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service