Skip to Content
Merck
All Photos(1)

Key Documents

447609

Sigma-Aldrich

Hexamethyldigermanium(IV)

technical grade

Synonym(s):

1,1,1,2,2,2-Hexamethyldigermane, Hexamethyldigermane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3GeGe(CH3)3
CAS Number:
Molecular Weight:
235.49
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

grade

technical grade

form

liquid

reaction suitability

core: germanium

refractive index

n20/D 1.456 (lit.)

bp

137-138 °C (lit.)

mp

−40 °C (lit.)

density

1.175 g/mL at 25 °C (lit.)

SMILES string

C[Ge](C)(C)[Ge](C)(C)C

InChI

1S/C6H18Ge2/c1-7(2,3)8(4,5)6/h1-6H3

InChI key

HLEHOFVKHRHBQI-UHFFFAOYSA-N

General description

Hexamethyldigermanium (IV) (HMGe) is a versatile compound used in the synthesis of a variety of organogermanium compounds, drugs, and catalysts.

Application

Hexamethyldigermanium (IV) used:
  • To synthesize digermanylated N-heterocycle compounds.
  • For the C–H germylation of nonterminal alkenes and aryl iodides.
  • To synthesize Ge Nanowires, Ge composites with other metals using low pressure chemical vapor deposition(LPCVD).

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

57.2 °F - closed cup

Flash Point(C)

14 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pd-Catalyzed Hydroxyl-Directed Cascade Hydroarylation/C?H Germylation of Nonterminal Alkenes and Aryl Iodides
Chun-Yan Wu, et al.},
The Journal of Organic Chemistry, 87, 9184-9196 (2022)
Formation of Cu 1-x Ge x Nanoplatelets Using LPCVD of Ge2Me6 or Ge2Me6/Et4 Pb Mixture
V. Drinek, et al.
NANO, 10, 1550061-1550061 (2015)
Palladium-Catalyzed Disilylation and Digermanylation of Alkene Tethered Aryl Halides: Direct Access to Versatile Silylated and Germanylated Heterocycles
Marco Wollenburg, et al.
Organic Letters, 22, 3679-3683 (2020)
Journal of the Chemical Society. Chemical Communications, 1236-1236 (1990)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service