Skip to Content
Merck
All Photos(1)

Key Documents

208450

Sigma-Aldrich

Germanium(IV) chloride

99.99% trace metals basis

Synonym(s):

Germanium tetrachloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
GeCl4
CAS Number:
Molecular Weight:
214.45
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

76 mmHg ( 20 °C)

Assay

99.99% trace metals basis

form

liquid

reaction suitability

reagent type: catalyst
core: germanium

impurities

≤150.0 ppm Trace Metal Analysis

bp

83 °C (lit.)

mp

−49.5 °C (lit.)

density

1.844 g/mL at 25 °C

SMILES string

Cl[Ge](Cl)(Cl)Cl

InChI

1S/Cl4Ge/c1-5(2,3)4

InChI key

IEXRMSFAVATTJX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Germanium tetrachloride may be used in the microwave preparation of Ge2Cl6, a colorless crystalline material which was included in a further study of low-valent germanium compounds.GeCl4 may be used in the plasma enhanced vapor deposition of germanium chalcogenide thin films. GeCl4 in combination with triphenylphosphine may be applied for the reduction of a number of a-bromo carboxylic acid derivatives.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 3 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Does not sustain combustion.

Flash Point(C)

Does not sustain combustion.

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Triphenylphosphine/germanium(IV) chloride combination: A new agent for the reduction of a-bromo carboxylic acid derivatives
Kagoshima H, et al.
Tetrahedron Letters, 39(10), 1203-1206 (1998)
GeS2 and GeSe2 PECVD from GeCl4 and Various Chalcogenide Precursors.
Whitham PJ, et al.
Plasma Chemistry and Plasma Processing, 34(4), 755-766 (2014)
Zheng Gong et al.
Physical chemistry chemical physics : PCCP, 19(36), 24513-24521 (2017-07-20)
Spinel-phase Zn
Ian R. Beattie et al.
Inorganic chemistry, 37(23), 6032-6034 (2001-10-24)
The Raman spectrum of Ge(2)Cl(6) shows close analogies with that of the isoelectronic [Ga(2)Cl(6)](2)(-). A band at 262 cm(-)(1) is assigned as principally a Ge-Ge stretching mode. The compound is very moisture-sensitive and decomposes readily in vacuo to give GeCl(4)
Murat Kaya et al.
Talanta, 84(1), 122-126 (2011-02-15)
A new chloride generation system was designed for the direct, sensitive, rapid and accurate determination of the total germanium in complex matrices. It was aimed to improve the detection limit of chloride generation technique by increasing the vapor pressure of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service