Skip to Content
Merck
All Photos(2)

Documents

429392

Sigma-Aldrich

Cesium bromide

AnhydroBeads, −10 mesh, 99.999% trace metals basis

Synonym(s):

Cesium monobromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CsBr
CAS Number:
Molecular Weight:
212.81
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

product line

AnhydroBeads

Quality Level

Assay

99.999% trace metals basis

form

beads

impurities

≤15.0 ppm Trace Metal Analysis

particle size

−10 mesh

mp

636 °C (lit.)

density

4.44 g/mL at 25 °C (lit.)

SMILES string

[Br-].[Cs+]

InChI

1S/BrH.Cs/h1H;/q;+1/p-1

InChI key

LYQFWZFBNBDLEO-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Legal Information

AnhydroBeads is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Caesium-binding sites in the gramicidin pore.
D A Doyle et al.
Biochemical Society transactions, 22(4), 1043-1045 (1994-11-01)
Matthieu P Wintergerst et al.
Journal of the American Chemical Society, 130(12), 4129-4139 (2008-03-04)
Solvent-extraction studies provide confirming evidence that meso-octamethylcalix[4]pyrrole acts as an ion-pair receptor for cesium chloride and cesium bromide in nitrobenzene solution. The stoichiometry of the interaction under extraction conditions from water to nitrobenzene was determined from plots of the cesium
Peng Pan et al.
Analytical chemistry, 75(20), 5468-5474 (2004-01-09)
Solutions consisting of protein and small molecule mixtures have been subjected to electrospray ionization to study the influence of small molecule/cation components at high concentrations on the electrospray responses of proteins. Emphasis was placed on solutions consisting of equal parts
N Kurata et al.
Radiation protection dosimetry, 119(1-4), 398-401 (2006-06-01)
CsBr phosphor ceramics doped with different luminescence centres such as In2O3, Eu2O3, EuCl3, SmCl3, TbCl3, GdCl3 or NdCl3 as the candidate for a new optically-stimulable phosphor for medical X-ray imaging sensor were prepared using a conventional ceramic fabrication process. It
Wen-Chen Zheng et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 64(3), 628-630 (2006-01-03)
The complete high-order perturbation formula of g factor, including not only the widely used crystal-field (CF) mechanism, but also the neglected change-transfer (CT) mechanism in the CF theory, is established for a 3d8 ion in cubic octahedral site. From the

Articles

Colloidal quantum dots (CQDs) are semiconducting crystals of only a few nanometers (ca. 2–12 nm) coated with ligand/surfactant molecules to help prevent agglomeration.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service