Skip to Content
Merck
All Photos(1)

Key Documents

554359

Sigma-Aldrich

Lead(II) iodide

AnhydroBeads, −10 mesh, 99.999% trace metals basis

Synonym(s):

Lead diiodide, Lead diiodide (PbI2), Lead iodide, Plumbous iodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
PbI2
CAS Number:
Molecular Weight:
461.01
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

product line

AnhydroBeads

Assay

99.999% trace metals basis

impurities

≤15.0 ppm Trace Metal Analysis

particle size

−10 mesh

bp

954 °C (lit.)

mp

402 °C (lit.)

density

6.16 g/mL at 25 °C (lit.)

SMILES string

I[PbH2]I

InChI

1S/2HI.Pb/h2*1H;/q;;+2/p-2

InChI key

RQQRAHKHDFPBMC-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Lead(II) iodide is a wide band gap(2.32 eV)semiconductor material with unique properties such as high resistivity, a widetemperature range (−200 °C up to +130 °C), and good chemical stability.

Application

Lead(II)iodide can be used as a starting material to prepare:

  • Polycrystalline α-FAPbI3 thin films by solution processing method. These polycrystalline thin films are applicable as photodetectors.
  • Organic/inorganic hybrid 2D perovskite materials, applicable in solar cells.
It can also be used to fabricate a gamma-ray detector at room temperature.

Legal Information

AnhydroBeads is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 1A - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Dye-sensitized solar cells as a promising, low-cost photovoltaic technology.

Dye-sensitized solar cells as a promising, low-cost photovoltaic technology.

Dye-sensitized solar cells as a promising, low-cost photovoltaic technology.

Dye-sensitized solar cells as a promising, low-cost photovoltaic technology.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service