Skip to Content
Merck
All Photos(3)

Key Documents

416320

Sigma-Aldrich

Poly(methyl vinyl ether-alt-maleic anhydride)

average Mw ~1,080,000, average Mn ~311,000

Synonym(s):

maleated copolymer, maleated polymer, maleic copolymer

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C4H2O3 · C3H6O)n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

powder

Quality Level

mol wt

average Mn ~311,000
average Mw ~1,080,000

impurities

<2% benzene

solubility

H2O: soluble (upon hydrolysis)
organic solvents: soluble

SMILES string

O1C(=O)C=CC1=O.O(C)C=C

InChI

1S/C4H2O3.C3H6O/c5-3-1-2-4(6)7-3;1-3-4-2/h1-2H;3H,1H2,2H3

InChI key

UPBDXRPQPOWRKR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Poly(methyl vinyl ether-alt-maleic anhydride) P(MVE-MA) can be used:
  • To prepare antimicrobial hydrogels with good mechanical strength, adhesiveness to the skin, and excellent swelling ability. For example, autoclaved mixtures of P(MVE-MA) and PVA (polyvinyl alcohol) can inhibit bacteria growth against Staphylococcus aureus.
  • As an electrolyte additive for stabilizing the interface of high-voltage layered Li-rich oxides (LLROs). P(MVE-MA) can absorb more H2O from the electrolyte and suppress the oxidative decomposition of the electrolyte.
  • To functionalize electrospun PVA nanofibers for protein adsorption. These modified nanofibrous membranes can be used for protein purification.
  • To synthesize separator membranes with good ionic conductivity. These membranes keep the positive and negative electrodes apart and prevent electrical short circuits.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1A - Eye Irrit. 2 - Muta. 1B - Skin Irrit. 2 - STOT RE 2

Target Organs

Blood

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Suru Mankodi et al.
American journal of dentistry, 24 Spec No A, 21A-27A (2012-01-06)
To investigate the efficacy of a dentifrice containing 0.3% triclosan, 2.0% PVM/MA copolymer, 0.243% sodium fluoride, and specially-designed silica in controlling established dental plaque and gingivitis. Qualifying adult male and female subjects from the West Palm Beach, Florida area were
Xuejun Liu et al.
Journal of dentistry, 39(4), 293-301 (2010-11-09)
To study the effectiveness of a dentifrice containing polymethyl vinyl ether-maleic acid (PVM/MA) copolymer in a silica base in occluding dentin tubules for treatment of dentin sensitivity. Thirty-two human dentin discs were divided into two groups and brushed in the
Superior management of plaque and gingivitis through the use of a triclosan/copolymer dentifrice.
Sebastian Ciancio et al.
The Journal of clinical dentistry, 21(4), 93-95 (2011-01-29)
Sebastian G Ciancio
Compendium of continuing education in dentistry (Jamesburg, N.J. : 1995), 32(1), 70-76 (2011-04-06)
This review summarizes research that has assessed the effectiveness of various antimicrobial-containing dentifrices in preventing and/or reducing a number of oral health problems facing our patients today. The results of these studies indicate that, when compared with a conventional fluoride
A dentifrice with multiple benefits.
Franklin García-Godoy
American journal of dentistry, 24 Spec No A, 2A-2A (2012-01-06)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service