Skip to Content
Merck
All Photos(3)

Key Documents

416339

Sigma-Aldrich

Poly(methyl vinyl ether-alt-maleic anhydride)

average Mw ~216,000, average Mn ~80,000

Synonym(s):

maleated copolymer, maleated polymer, maleic copolymer

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C4H2O3 · C3H6O)n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

powder

Quality Level

mol wt

average Mn ~80,000
average Mw ~216,000

impurities

<2% benzene

solubility

H2O: soluble (upon hydrolysis)
organic solvents: soluble

SMILES string

O1C(=O)C=CC1=O.O(C)C=C

InChI

1S/C4H2O3.C3H6O/c5-3-1-2-4(6)7-3;1-3-4-2/h1-2H;3H,1H2,2H3

InChI key

UPBDXRPQPOWRKR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Film former, dispersant, emulsifier, thickener, binder, flocculant and complexing agent.

Features and Benefits

Reacts with alcohols, amines and epoxides to form derivatives or cross-linked polymers. Readily derivatized. Forms iodine complex.

Physical form

Alternating copolymer.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1A - Eye Irrit. 2 - Muta. 1B - Skin Irrit. 2 - STOT RE 2

Target Organs

Blood

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Suru Mankodi et al.
American journal of dentistry, 24 Spec No A, 21A-27A (2012-01-06)
To investigate the efficacy of a dentifrice containing 0.3% triclosan, 2.0% PVM/MA copolymer, 0.243% sodium fluoride, and specially-designed silica in controlling established dental plaque and gingivitis. Qualifying adult male and female subjects from the West Palm Beach, Florida area were
Sebastian G Ciancio
Compendium of continuing education in dentistry (Jamesburg, N.J. : 1995), 32(1), 70-76 (2011-04-06)
This review summarizes research that has assessed the effectiveness of various antimicrobial-containing dentifrices in preventing and/or reducing a number of oral health problems facing our patients today. The results of these studies indicate that, when compared with a conventional fluoride
Katarzyna Migalska et al.
Pharmaceutical research, 28(8), 1919-1930 (2011-03-26)
To assess the feasibility of transdermal macromolecule delivery using novel laser-engineered dissolving microneedles (MNs) prepared from aqueous blends of 20% w/w poly(methylvinylether maleic anhydride) (PMVE/MA) in vitro and in vivo. Micromoulding was employed to prepare insulin-loaded MNs from aqueous blends
A dentifrice with multiple benefits.
Franklin García-Godoy
American journal of dentistry, 24 Spec No A, 2A-2A (2012-01-06)
R M Khashaba et al.
International endodontic journal, 44(4), 342-352 (2011-01-29)
To evaluate the histopathologic biocompatibility of two new calcium phosphate-based sealers (CPS-1 & CPS-2) with a commercially available calcium hydroxide-based sealer (Acroseal). Polyethylene tubes were filled with freshly mixed sealers and implanted subcutaneously in the dorsal connective tissue of rats.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service