Skip to Content
Merck
All Photos(1)

Key Documents

409073

Sigma-Aldrich

Trimethylolpropane ethoxylate triacrylate

average Mn ~428

Synonym(s):

Ethoxylated trimethylolpropane triacrylate, Polyethylene glycol trimethylolpropane ether triacrylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[H2C=CHCO2(CH2CH2O)nCH2]3CC2H5
CAS Number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

liquid

mol wt

average Mn ~428

refractive index

n20/D 1.471 (lit.)

bp

157 °C (lit.)

density

1.11 g/mL at 25 °C

SMILES string

CCC(COCCOC(=O)C=C)(COCCOC(=O)C=C)COCCOC(=O)C=C

InChI

1S/C21H32O9/c1-5-18(22)28-12-9-25-15-21(8-4,16-26-10-13-29-19(23)6-2)17-27-11-14-30-20(24)7-3/h5-7H,1-3,8-17H2,4H3

InChI key

MTPIZGPBYCHTGQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Trimethylolpropane ethoxylate triacrylate belongs to the class of monomers known as acrylate esters. It exhibits low viscosity and high reactivity due presence of multiple acrylate functional groups which allows rapid polymerization and cross-linking. This monomer is widely used to prepare UV-curable resins for 3D printing, optical coatings for display devices, and dental composite restorative materials. It also possesses inherent hydrophobic characteristics suitable for applications where water resistance or moisture barrier properties are required.

Application

Trimethylolpropane ethoxylate triacrylate can be used:
  • As a cross-linking agent or a polymerizable monomer in the preparation of gel polymer electrolyte for Lithium-ion batteries. It enhances the thermal stability, ionic conductivity, and mechanical strength of the polymer.
  • As a monomer to prepare UV-curable resins via vat photopolymerization, for stereolithography (SLA) and digital light processing (DLP). These techniques have potential applications in the 3D printing of medical devices, soft robots, dental implants, Optical adhesives, and microfluidic devices.
  • To synthesize inverse micropatterned photonic crystal (PhC) films via photopolymerization for the detection of alcohol content in water.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Skin Sens. 1B

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High-Performance and Highly Safe Solvate Ionic Liquid-Based Gel Polymer Electrolyte by Rapid UV-Curing for Lithium-Ion Batteries
Xinzhu Gao, et al.
ACS Applied Materials & Interfaces, 14, 43397-43406 (2022)
Fabrication Technique of Micropatterned Inverse Photonic Crystal Films
Matin Ashurov, et al.
Crystal Research and Technology, 58, 2300224-2300224 (2023)
Recent trends in advanced photoinitiators for vat photopolymerization 3D printing
Yinyin Bao
Macromolecular Rapid Communications, 43, 2200202-2200202 (2022)
Sophia Katharine Laney et al.
ACS nano, 14(9), 12091-12100 (2020-08-20)
Periodic nanotube arrays render enhanced functional properties through their interaction with light and matter, but to reach optimal performance for technologically prominent applications, such as wettability or photonics, structural fine-tuning is essential. Nonetheless, a universal and scalable method providing independent

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service