Skip to Content
Merck
All Photos(1)

Documents

246832

Sigma-Aldrich

Tri(propylene glycol) diacrylate, mixture of isomers

contains MEHQ and HQ as inhibitors, technical grade

Synonym(s):

TRPGDA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHCO(OC3H6)3O2CCH=CH2
CAS Number:
Molecular Weight:
300.35
EC Number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

grade

technical grade

Quality Level

vapor density

>1 (vs air)

vapor pressure

<0.01 mmHg ( 20 °C)

form

liquid

contains

150-220 ppm monomethyl ether hydroquinone and hydroquinone (total inhibitor)

refractive index

n20/D 1.45 (lit.)

density

1.03 g/mL at 25 °C (lit.)

SMILES string

CC(COC(C)COC(=O)C=C)OCC(C)OC(=O)C=C

InChI

1S/C15H24O6/c1-6-14(16)20-12(4)9-18-8-11(3)19-10-13(5)21-15(17)7-2/h6-7,11-13H,1-2,8-10H2,3-5H3

InChI key

ZDQNWDNMNKSMHI-UHFFFAOYSA-N

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

307.4 °F - closed cup

Flash Point(C)

153 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bor-Shiunn Lee et al.
Journal of endodontics, 34(5), 594-598 (2008-04-26)
We have developed a visible-light curable urethane-acrylate/tripropylene glycol diacrylate (UA/TPGDA) oligomer to serve as a root canal sealer and a zinc oxide/thermoplastic polyurethane (ZnO/TPU) composite to serve as a root canal obturation material. The purpose of this study was to
L A Nylander-French et al.
Toxicologic pathology, 26(4), 476-483 (1998-08-26)
The toxicity of the esters of acrylic acid are poorly understood even though significant human exposure occurs. To conduct rapid comparative short-term bioassays, we used the Tg.AC (v-Ha-ras) transgenic mouse model to determine the toxicity and potential carcinogenicity of tripropylene
Daniel J Duffy et al.
Journal of the American Chemical Society, 124(28), 8290-8296 (2002-07-11)
A model system for the characterization of molecular recognition events in molecularly imprinted polymers (MIPs) is presented. The use of a biologically inspired, three-point hydrogen-bonding motif and a thin film polymeric matrix allows for pre- and post-polymerization binding properties to
R R Tice et al.
Environmental and molecular mutagenesis, 29(3), 240-249 (1997-01-01)
Acrylates may be polymerized to stable surface coatings (paints, lacquers, inks, etc.) by alkylation via the Michaelis-type addition reaction. Thus, acrylates have an inherent potential as electrophiles to be genotoxic, limited in their biological activity by their physicochemical properties. To
L A Nylander-French et al.
In vitro cellular & developmental biology. Animal, 36(9), 611-616 (2001-02-24)
The potential for occupational exposure to the esters of acrylic acid (acrylates) is considerable, and, thus, requires a greater understanding of the their toxicity. Confluent (70-90%) cultures of normal human epidermal keratinocytes (NHEK), dermal fibroblasts (NHDF). or bronchial epithelium (NHBE)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service