Skip to Content
Merck
All Photos(1)

Key Documents

246808

Sigma-Aldrich

Trimethylolpropane triacrylate

contains monomethyl ether hydroquinone as inhibitor, technical grade

Synonym(s):

TMPTA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(H2C=CHCO2CH2)3CC2H5
CAS Number:
Molecular Weight:
296.32
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

grade

technical grade

vapor density

>1 (vs air)

vapor pressure

<0.01 mmHg ( 20 °C)

contains

monomethyl ether hydroquinone as inhibitor

refractive index

n20/D 1.474 (lit.)

density

1.1 g/mL at 25 °C (lit.)

SMILES string

CCC(COC(=O)C=C)(COC(=O)C=C)COC(=O)C=C

InChI

1S/C15H20O6/c1-5-12(16)19-9-15(8-4,10-20-13(17)6-2)11-21-14(18)7-3/h5-7H,1-3,8-11H2,4H3

InChI key

DAKWPKUUDNSNPN-UHFFFAOYSA-N

General description

Trimethylolpropanetriacrylate (TMPTA) is a tri-functional monomer that is used in the productionof various types of polymers with a wide range of applications. The most commonuse of trimethylolpropane triacrylate is in the production of UV-curedcoatings, inks, and adhesives, resins, photosensitive materials, andsuperabsorbent materials. Additionally, TMPTA-based polymers are utilized in theproduction of thermosetting resins, which are used in the production ofcomposites, laminates, and coatings. TMPTA can also be copolymerized with othermonomers to tailor the properties of the resulting polymers to meet specificrequirements.

Application

Trimethylolpropane triacrylate can be used as a starting material in the preparation of:
  • Poly(trimethylolpropane triacrylate/Al(PTMPTA/Al) nanocomposites by in situ solution polymerization.
  • Cross-linkable acrylamide/crotonic acid water retainer hydrogels.
  • Polymer electrolyte for dye-sensitized solar cells.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

382.1 °F - closed cup

Flash Point(C)

194.5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Surface-tension-induced synthesis of complex particles using confined polymeric fluids.
Chang-Hyung Choi et al.
Angewandte Chemie (International ed. in English), 49(42), 7748-7752 (2010-09-04)
Yongsheng Gao et al.
Biomacromolecules, 17(11), 3640-3647 (2016-09-20)
A successful polymeric gene delivery vector is denoted by both transfection efficiency and biocompatibility. However, the existing vectors with combined high efficacy and minimal toxicity still fall short. The most widely used polyethylene imine (PEI), polyamidoamine (PAMAM) and poly(dimethylaminoethyl methacrylate)
Xiaoying Zhao et al.
Polymers, 11(3) (2019-04-10)
Poly(3-hydroxybutyrate-co-3-hydroxyvalerate) (PHBV) is a promising bio-based and biodegradable thermoplastic with restricted industrial applications due to its brittleness and poor processability. Natural rubber (NR) has been used as a toughening agent, but further physical improvements are desired. In this study, rubber
Keng-Chi Cho et al.
Optics express, 19(12), 11732-11739 (2011-07-01)
In this study, the intensity of two-photon excited fluorescence (TPEF) of xanthene dye, Rose Bengal (RB), was significantly enhanced via bovine serum albumin (BSA) microstructures fabricated by the two-photon crosslinking (TPC) technique. The RB was utilized as the photoactivator in
G Gunadi et al.
Dental materials : official publication of the Academy of Dental Materials, 13(1), 7-12 (1997-01-01)
The purpose of this study was to prepare a light-cured adhesive applicable for orthodontics by mixing monomers and a polymerized reactive organic composite filler (prepolymerized trimethylolpropane trimethacrylate-filler, TMPT-filler). The monomer component was a mixture of 3.0 wt% 2-hydroxy-3-(2-naphthoxy)propyl methacrylate (HNPM)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service