Skip to Content
Merck
All Photos(2)

Key Documents

407801

Sigma-Aldrich

6-Methyl-1,3,5-triazine-2,4-diamine

98%

Synonym(s):

2,4-Diamino-6-methyl-1,3,5-triazine, Acetoguanamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H7N5
CAS Number:
Molecular Weight:
125.13
Beilstein:
118348
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

powder

mp

274-276 °C (lit.)

SMILES string

Cc1nc(N)nc(N)n1

InChI

1S/C4H7N5/c1-2-7-3(5)9-4(6)8-2/h1H3,(H4,5,6,7,8,9)

InChI key

NJYZCEFQAIUHSD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

6-Methyl-1,3,5-triazine-2,4-diamine butane-1,4-diol monosolvate crystallizes with 6-methyl-1,3,5-triazine-2,4-diamine (DMT) and a molecule of butane-1,4-diol in the asymmetric unit. The DMT molecules form ribbons having centrosymmetric dimer motifs between DMT molecules along the c-axis. DMT and thymine forms a new metastable bicomponent hydrogel, which cage a number of organic solvents.

Application

6-Methyl-1,3,5-triazine-2,4-diamine may be used in the synthesis of s-Bodipy-triazine (BODIPY = 4,4-difluoro-4-borata-3a-azonia-4a-aza-s-indacene). It is a symmetric fluorescent sensor for Cu2+.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Metastability in a bi-component hydrogel of thymine and 6-methyl-1, 3, 5-triazine-2, 4-diamine: ultrasound induced vs. thermo gelation.
Roy B, et al.
Soft Matter, 8(8), 2366-2369 (2012)
A symmetric and selective fluorescent Cu (II) sensor based on bodipy and s-triazine.
Kursunlu AN, et al.
Journal of Luminescence, 149, 215-220 (2014)
Rajni M Bhardwaj et al.
Acta crystallographica. Section E, Structure reports online, 68(Pt 12), o3377-o3377 (2013-03-12)
The title co-crystal, C4H7N5·C4H10O2, crystallizes with one mol-ecule of 6-methyl-1,3,5-triazine-2,4-diamine (DMT) and one mol-ecule of butane-1,4-diol in the asymmetric unit. The DMT mol-ecules form ribbons involving centrosymmetric R2(2)(8) dimer motifs between DMT mol-ecules along the c-axis direction. These ribbons are
Kun Zhang et al.
Analytica chimica acta, 1144, 34-42 (2021-01-18)
Herein, we report our strategy to develop the efficient chemosensor and real-time monitoring technique for cyanuric chloride (TCT) detection. A luminescent macrocyclic mononuclear Sm(III) complex Sm-2k bearing with two dynamic imine bonds has been constructed via the template synthesis between

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service