D26202
3,5-Diamino-1,2,4-triazole
98%
Synonym(s):
Guanazole
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C2H5N5
CAS Number:
Molecular Weight:
99.09
Beilstein:
112467
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
98%
form
solid
mp
202-205 °C (lit.)
SMILES string
Nc1n[nH]c(N)n1
InChI
1S/C2H5N5/c3-1-5-2(4)7-6-1/h(H5,3,4,5,6,7)
InChI key
PKWIYNIDEDLDCJ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
3,5-Diamino-1,2,4-triazole, also known as Guanazole is a heterocyclic compound and is commonly used as a building block in the synthesis of gallium complexes of tert-butyl-substituted acyclic and cyclic compounds. It is also being investigated as a corrosion inhibitor for copper.
Application
Inhibitor of DNA synthesis.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2 - Repr. 2 - STOT RE 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
[Pharmacokinetics of antitumor drugs].
V A Filov et al.
Voprosy onkologii, 29(7), 3-13 (1983-01-01)
R G Hards et al.
Journal of cellular physiology, 106(2), 309-319 (1981-02-01)
We describe the isolation and characterization of a Chinese hamster ovary cell line selected for resistance to N-carbamoyloxyurea. Using the mammalian cell permeabilization assay developed in our laboratory, a detailed analysis of the target enzyme, ribonucleotide reductase (EC 1.17.4.1), was
Inhibition of ribonucleotide reductases encoded by herpes simplex viruses.
T Spector
Pharmacology & therapeutics, 31(3), 295-302 (1985-01-01)
Studies on the selective toxicity of guanazole in mice.
C Dave et al.
European journal of cancer, 14(1), 33-40 (1978-01-01)
C Loganathan et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 173, 837-842 (2016-11-07)
Detection of a chromogenic insecticide, malachite green (MG) using 3,5-diamino-1,2,4-triazole capped gold nanoparticles (DAT-AuNPs) by both naked eye and spectrophotometry was described in this paper. The DAT-AuNPs were prepared by wet chemical method and show absorption maximum at 518nm. The
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service