Skip to Content
Merck
All Photos(3)

Key Documents

D5758

Sigma-Aldrich

Diethyl pyrocarbonate

96% (NT)

Synonym(s):

DEP, DEPC, Diethyl dicarbonate, Diethyl oxydiformate, Ethoxyformic acid anhydride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O(COOC2H5)2
CAS Number:
Molecular Weight:
162.14
Beilstein:
637031
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic (microbial fermentation/ cell culture)

Quality Level

Assay

96% (NT)

form

liquid

refractive index

n20/D 1.398 (lit.)

bp

93-94 °C/18 mmHg (lit.)

density

1.101 g/mL at 25 °C (lit.)

shipped in

wet ice

storage temp.

2-8°C

SMILES string

CCOC(=O)OC(=O)OCC

InChI

1S/C6H10O5/c1-3-9-5(7)11-6(8)10-4-2/h3-4H2,1-2H3

InChI key

FFYPMLJYZAEMQB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diethyl Pyrocarbonate (DEPC) is a chemical use to inactivate RNase enzymes and is sensitive to moisture and pH. It decomposes at 155°C, to ethanol and carbon dioxide in aqueous solution. DEPC is also sensitive to ammonia, which causes decomposition to urethane, a possible carcinogen.

Application

Diethyl pyrocarbonate has been used in PCR reaction for treating deionized water, which reduces the risk of RNA being degraded by RNases. It is also used for Dot blot hybridization, to dilute total RNA isolated from different micro-organisms.
Modification reagent for His and Tyr residues in proteins. Robust probe for structural disruptions in dsDNA, reacting with fully or partially unstacked bases.

Biochem/physiol Actions

DEPC is a potent nuclease inhibitor.0.1% solution of DEPC is used to inactive RNase.
It inhibits the ryanodine binding to ryanodine/Ca2+ receptor channels in skeletal muscle in a dose and time dependent manner and increases the Ca2+ permeability of SR vesicles.
Inactivates RNase in solution at about 0.1% (v/v), thus protecting RNA against degradation.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

156.2 °F - closed cup

Flash Point(C)

69 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

H Perry-O'Keefe et al.
Journal of applied microbiology, 90(2), 180-189 (2001-02-13)
A method for rapid and simultaneous detection, identification and enumeration of specific micro-organisms using Peptide Nucleic Acid (PNA) probes is presented. The method is based on a membrane filtration technique. The membrane filter was incubated for a short period of
V Shoshan-Barmatz et al.
The Biochemical journal, 299 ( Pt 1), 177-181 (1994-04-01)
Exposure of junctional sarcoplasmic reticulum (SR) membranes or purified ryanodine receptor to the histidine-specific reagent diethyl pyrocarbonate (DEPC) led to concentration- and time-dependent inactivation of ryanodine binding. The pH-dependence of the inactivation of ryanodine binding by DEPC and the reversal
Properties of mouse globin messenger ribonucleic acid and its preparation in milligram quantities.
R Williamson et al.
Biochemistry, 10(16), 3014-3021 (1971-08-03)
Melina Papalampropoulou-Tsiridou et al.
Frontiers in cellular neuroscience, 14, 103-103 (2020-06-09)
Injury and inflammation cause tissue acidosis, which is a common feature of various painful conditions. Acid-Sensing Ion channels (ASICs) are amongst the main excitatory channels activated by extracellular protons and expressed in the nervous system. Six transcripts of ASIC subunits
E C Williamson et al.
Journal of clinical microbiology, 39(1), 47-50 (2001-01-04)
Respiratory tract colonization with Scedosporium apiospermum in patients with chronic suppurative lung disease is a significant concern for lung transplantation candidates, since Scedosporium infections occurring posttransplantation are usually untreatable. Up to 10% of patients with cystic fibrosis attending our respiratory

Articles

Nuclease-free water (or PCR water) is needed in molecular biology to avoid sample degradation during PCR, sequencing, etc. Ultrafiltration provides a quicker, safer, and more sustainable alternative to DEPC treatment when preparing RNase-free and DNase-free water.

Nuclease-free water (or PCR water) is needed in molecular biology to avoid sample degradation during PCR, sequencing, etc. Ultrafiltration provides a quicker, safer, and more sustainable alternative to DEPC treatment when preparing RNase-free and DNase-free water.

Nuclease-free water (or PCR water) is needed in molecular biology to avoid sample degradation during PCR, sequencing, etc. Ultrafiltration provides a quicker, safer, and more sustainable alternative to DEPC treatment when preparing RNase-free and DNase-free water.

Nuclease-free water (or PCR water) is needed in molecular biology to avoid sample degradation during PCR, sequencing, etc. Ultrafiltration provides a quicker, safer, and more sustainable alternative to DEPC treatment when preparing RNase-free and DNase-free water.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service