Skip to Content
Merck
All Photos(1)

Key Documents

392723

Sigma-Aldrich

(DHQ)2PHAL

≥95%

Synonym(s):

Hydroquinine 1,4-phthalazinediyl diether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C48H54N6O4
CAS Number:
Molecular Weight:
778.98
Beilstein:
5475677
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥95%

optical activity

[α]22/D +336°, c = 1.2 in methanol

mp

178 °C (dec.) (lit.)

SMILES string

CC[C@@H]1CN2CCC1CC2[C@H](Oc3nnc(O[C@@H](C4CC5CCN4C[C@@H]5CC)c6ccnc7ccc(OC)cc67)c8ccccc38)c9ccnc%10ccc(OC)cc9%10

InChI

1S/C48H54N6O4/c1-5-29-27-53-21-17-31(29)23-43(53)45(35-15-19-49-41-13-11-33(55-3)25-39(35)41)57-47-37-9-7-8-10-38(37)48(52-51-47)58-46(44-24-32-18-22-54(44)28-30(32)6-2)36-16-20-50-42-14-12-34(56-4)26-40(36)42/h7-16,19-20,25-26,29-32,43-46H,5-6,17-18,21-24,27-28H2,1-4H3/t29-,30+,31-,32+,43-,44-,45-,46-/m1/s1

InChI key

YUCBLVFHJWOYDN-PPIALRKJSA-N

Looking for similar products? Visit Product Comparison Guide

General description

DHQ)2PHAL is a modified cinchona alkaloid.

Application

(DHQ)2PHAL may be used in the following processes:
  • As a catalyst Asymmetric and chemoselective N-allylic alkylation of indoles with Morita-Baylis-Hillman carbonates to form pyrrolo[1,2-a]indole and pyrrolo[3,2,1-ij]quinoline derivatives.
  • As a ligand for the osmium catalyzed-Sharpless asymmetric dihydroxylation step of (S)-a-benzoyloxy carboxylic acids multistep synthesis.
  • As a ligand for the carbamate based asymmetric aminohydroxylation of styrene derivatives to form N-carbamate protected R-arylglycinols.
Ligand for Sharpless Asymmetric Dihydroxylation employed in an enantioselective preparation of syn-3,5-hydroxycarboxylates from (E,E)- or (E,Z)-1,3-dienoates.

Physical form

Asymmetric dihydroxylation of unsaturated esters with trisubstituted C=C bonds.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

From styrenes to enantiopure a-arylglycines in two steps.
Reddy KL and Sharpless KB.
Journal of the American Chemical Society, 120(6), 1207-1217 (1990)
Kapferer, Tobias; Brueckner, Reinhard
European Journal of Organic Chemistry, 9, 2119-2133 (2006)
Chemoselective Asymmetric N-Allylic Alkylation of Indoles with Morita-Baylis-Hillman Carbonates.
Cui H-L, et al.
Angewandte Chemie (International Edition in English), 48(31), 5737-5740 (2009)
Regioselective Oxidative Cleavage of Benzylidene Acetals: Synthesis of a-and ?-Benzoyloxy Carboxylic Acids.
Kumar PS, et al.
Angewandte Chemie (International Edition in English), 49(4), 804-807 (2010)
Md Moinuddin Ahmed et al.
The Journal of organic chemistry, 71(20), 7741-7746 (2006-09-26)
The enantioselective syntheses of several protected 4-substituted syn-3,5-dihydroxy carboxylic esters have been achieved from the corresponding achiral (E,E)- or (E,Z)-1,3-dienoates. The route relies upon an enantio- and regioselective Sharpless dihydroxylation and a palladium-catalyzed reduction to form gamma-substituted delta-hydroxy-1-enoates. The resulting

Articles

Asymmetric phase-transfer catalysis (PTC) has been recognized as a “green” alternative to many homogeneous synthetic organic transformations, and has found widespread application. Synthetically modified cinchona alkaloids are typical chiral organocatalysts used in asymmetric PTC.

Asymmetric phase-transfer catalysis (PTC) has been recognized as a “green” alternative to many homogeneous synthetic organic transformations, and has found widespread application. Synthetically modified cinchona alkaloids are typical chiral organocatalysts used in asymmetric PTC.

Asymmetric phase-transfer catalysis (PTC) has been recognized as a “green” alternative to many homogeneous synthetic organic transformations, and has found widespread application. Synthetically modified cinchona alkaloids are typical chiral organocatalysts used in asymmetric PTC.

Asymmetric phase-transfer catalysis (PTC) has been recognized as a “green” alternative to many homogeneous synthetic organic transformations, and has found widespread application. Synthetically modified cinchona alkaloids are typical chiral organocatalysts used in asymmetric PTC.

Related Content

The Sharpless Lab pursues useful new reactivity and general methods for selectively controlling chemical reactions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service