Skip to Content
Merck
All Photos(1)

Key Documents

132225

Troger′s Base

98%

Synonym(s):

2,8-Dimethyl-6H,12H-5,11-methanodibenzo[b,f][1,5]diazocine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H18N2
CAS Number:
Molecular Weight:
250.34
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

powder

technique(s)

photometry: suitable

mp

133-136 °C (lit.)

SMILES string

Cc1ccc2[N@@H]3C[N@@H](Cc2c1)c4ccc(C)cc4C3

InChI

1S/C17H18N2/c1-12-3-5-16-14(7-12)9-18-11-19(16)10-15-8-13(2)4-6-17(15)18/h3-8H,9-11H2,1-2H3

InChI key

SXPSZIHEWFTLEQ-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R A Johnson et al.
Journal of medicinal chemistry, 36(21), 3202-3206 (1993-10-15)
The synthesis of 2,8-dimethyl-6H,12H-5,11-methanodibenzo[b,f][1,5]diazocine (Tröger's base) from p-toluidine and of two Tröger's base analogs from other anilines by reaction with hexamethylenetetramine in trifluoroacetic acid is described. 2,3,6,7-Tetrahydro-9-methyl-2,6-di-p-tolyl-1H,5H-pyrimido[5,6,1-ij] quinazoline is formed as a secondary product in the reaction of p-toluidine and
Tröger's-base-derived infinite co-ordination polymer microparticles.
You-Moon Jeon et al.
Small (Weinheim an der Bergstrasse, Germany), 5(1), 46-50 (2008-12-06)
J Elguero et al.
Magnetic resonance in chemistry : MRC, 43(8), 665-669 (2005-06-30)
The (1)H and (13)C NMR spectra of two stereoisomeric bis-Tröger's bases and four stereoisomeric tris-Tröger's bases asymmetrically substituted on the external aromatic rings were recorded and the corresponding signals assigned. The relative configuration of the stereogenic units has been unequivocally
Nicolas Claessens et al.
Journal of inorganic biochemistry, 101(7), 987-996 (2007-05-15)
Three stereoisomers of a Ru(II) complex bearing a chiral bis-phenanthroline Tröger's base analogue, TBphen2 (1), have been isolated from the reaction of the enantiomerically pure precursor complex Lambda- (or Delta-) cis-[Ru(phen)2(py)2]2+ (phen=1,10-phenanthroline, py=pyridine) with the racemic mixture of 1. Each
Xin Du et al.
Chemical communications (Cambridge, England), 46(6), 970-972 (2010-01-29)
An organic nanoporous polymer (BET surface area of 750 m(2) g(-1)) containing bicovalently-bonded Tröger's base as functional moieties in the network was synthesized, characterised, and further applied as a promising heterogeneous catalyst in the addition reaction of diethylzinc to aromatic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service