Skip to Content
Merck
All Photos(2)

Key Documents

379840

Sigma-Aldrich

Platinum(IV) chloride

≥99.99% trace metals basis

Synonym(s):

Platinum tetrachloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
PtCl4
CAS Number:
Molecular Weight:
336.89
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

≥99.99% trace metals basis

form

powder and chunks

reaction suitability

reagent type: catalyst
core: platinum

impurities

≤100.0 ppm Trace Metal Analysis

mp

370 °C (dec.) (lit.)

density

4.303 g/mL at 25 °C (lit.)

SMILES string

Cl[Pt](Cl)(Cl)Cl

InChI

1S/4ClH.Pt/h4*1H;/q;;;;+4/p-4

InChI key

FBEIPJNQGITEBL-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

General description

Platinum (IV)chloride is a highly oxidizing crystalline compound that can easily undergo reduction-oxidation reactions. It is widely used to synthesize Pt-based nanomaterials and catalysts.

Application

Platinum (IV) chloride can be used:
  • As a precursor to synthesize Pt nanoparticles for photocatalytic hydrogen generation.
  • As a starting material to prepare Pt/Al2O3 and Pt/ZrO2-SiO2 catalysts.
  • To fabricate an electrocatalyst for proton exchange fuel cells.
  • As a catalyst for the cyclization of arene−alkynesubstrates via intramolecular electrophilic hydroarylation.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hybrid metal-Cu2S nanostructures as efficient co-catalysts for photocatalytic hydrogen generation
Jeong-Hyun Park, et al.
Chemical Communications (Cambridge, England), 53, 3277-3280 (2017)
Third-body effects of native surfactants on Pt nanoparticle electrocatalysts in proton exchange fuel cells
Young-Hoon Chung, et al.
Chemical Communications (Cambridge, England), 51, 2968-2971 (2015)
Syngas production by carbon dioxide reforming of methane over Pt/Al2O3 and Pt/ZrO2-SiO2 catalysts
Ashvin L. Karemore, et al.
Chemical Engineering Science, 249, 117347-117347 (2022)
PtIV-Catalyzed Cyclization of Arene?Alkyne Substrates via Intramolecular Electrophilic Hydroarylation
Stefan J. Pastine, et al.},
Organic Letters, 5, 1055-1058 (2003)
Lu Zhao et al.
Zhejiang da xue xue bao. Yi xue ban = Journal of Zhejiang University. Medical sciences, 38(1), 59-66 (2009-03-03)
To construct a drug carrier and gene vector PEG-PEI-Pt. Polyethyleneglycol (PEG) was coupled to polyethylenimine (PEI 600) and platinum tetrachloride; PEG-PEI-Pt complex was formed in ethanol. The complex was characterized by XRD, UV-VIS and FT-IR and the DNA condensation was

Articles

Plasmonic nanoparticles have unique optical properties that can be tailored to suit a variety of applications in the biotechnology1–8 and electronics9–16 industries.

Plasmonic nanoparticles have unique optical properties that can be tailored to suit a variety of applications in the biotechnology1–8 and electronics9–16 industries.

Plasmonic nanoparticles have unique optical properties that can be tailored to suit a variety of applications in the biotechnology1–8 and electronics9–16 industries.

Plasmonic nanoparticles have unique optical properties that can be tailored to suit a variety of applications in the biotechnology1–8 and electronics9–16 industries.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service