Skip to Content
Merck
All Photos(2)

Key Documents

204021

Sigma-Aldrich

Ammonium hexachloroplatinate(IV)

99.995% trace metals basis

Synonym(s):

Ammonium platinic chloride, Platinum(IV)-ammonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)2PtCl6
CAS Number:
Molecular Weight:
443.87
EC Number:
MDL number:
UNSPSC Code:
12352302
eCl@ss:
38191314
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
solid

grade

for analytical purposes

Assay

99.995% trace metals basis

form

solid

impurities

≤55.0 ppm Trace Metal Analysis

density

3.07 g/mL at 25 °C (lit.)

SMILES string

[NH4+].[NH4+].Cl[Pt--](Cl)(Cl)(Cl)(Cl)Cl

InChI

1S/6ClH.2H3N.Pt/h6*1H;2*1H3;/q;;;;;;;;+4/p-4

InChI key

PCCGQTHFYHJATL-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Reacts with Cu(OAc)2 and H2S to form Cu2PtIIPt3IVS8 which has applications in diamagnetic materials and semiconductors.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R D Murdoch et al.
International archives of allergy and applied immunology, 77(4), 456-458 (1985-01-01)
Hooded Lister rats were sensitised to the halide salt of platinum ammonium tetrachloroplatinate(II)[(NH4)2PtCl4] in its conjugated form with ovalbumin. Sensitisation was achieved by intraperitoneal injection with Bordetella pertussis vaccine as adjuvant, followed 21 days later by a further injection in
Raghunandan Sharma et al.
ACS omega, 4(13), 15711-15720 (2019-10-02)
A water-assisted control of Pt nanoparticle size during a surfactant-free, microwave-assisted polyol synthesis of the carbon-supported platinum nanoparticles (Pt/C) in a mixture of ethylene glycol and water using (NH4)2PtCl6 as the Pt precursor is demonstrated. The particle size was tuned
A J Newman Taylor et al.
American journal of respiratory and critical care medicine, 160(2), 435-438 (1999-08-03)
The development of sensitization to inhaled allergens is determined by the interaction of multiple genetic and environmental influences. Occupational sensitization to low-molecular-weight chemicals allows a specific immunological response to an inhaled hapten to be studied in a well-defined population with
Allergy to the complex salts of platinum. A review of the literature and three case reports.
P Orbaek
Scandinavian journal of work, environment & health, 8(2), 141-145 (1982-06-01)
Sara Porro et al.
La Medicina del lavoro, 103(2), 123-129 (2012-05-25)
Occupational exposure to platinum salts may cause the onset of skin and respiratory disorders with an IgE-mediated allergic mechanism. The diagnosis of occupational asthma due to platinum salts was, in a small number of cases, achieved also via occupational specific

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service