Skip to Content
Merck
All Photos(2)

Key Documents

289175

Sigma-Aldrich

Neodymium(III) nitrate hexahydrate

99.9% trace metals basis

Synonym(s):

Neodymium nitrate hexahydrate, Neodymium trinitrate hexahydrate, Neodymium(3+) trinitrate hexahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Nd(NO3)3 · 6H2O
CAS Number:
Molecular Weight:
438.35
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.9% trace metals basis
form:
crystalline

Quality Level

Assay

99.9% trace metals basis

form

crystalline

reaction suitability

reagent type: catalyst
core: neodymium

impurities

≤1500.0 ppm Trace Rare Earth Analysis

SMILES string

O.O.O.O.O.O.[Nd+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/3NO3.Nd.6H2O/c3*2-1(3)4;;;;;;;/h;;;;6*1H2/q3*-1;+3;;;;;;

InChI key

VQVDTKCSDUNYBO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Neodymium(III) nitrate hexahydrate has a variety of uses such as:
  • fabrication of perovskite based solid oxide fuel cells.
  • synthesis of Nd3+ doped vanadium pentoxide nanostructure for potential usage in supercapacitors.
  • a catalyst for Friedlander synthesis of surface modified quinolones for application in medicinal chemistry.

Pictograms

Flame over circleExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A perovskite-type Nd 0.75 Sr 0.25 Co 0.8 Fe 0.2 O 3- delta cathode for advanced solid oxide fuel cells
Mulmi S and Thangadurai V
Chemical Communications (Cambridge, England), 55(26), 3713-3716 (2019)
Sudip Biswas et al.
Talanta, 206, 120176-120176 (2019-09-14)
Nd2O3 nanoparticle grafted graphene nanocomposite (NOGG) was synthesized by sonochemical treatment of an ethanolic dispersion of Nd2O3 nanoparticle and graphene. All the synthesized materials were characterized by XRD, FESEM, TEM, and BET. The NOGG has a high specific surface area
Asmae El Maangar et al.
Physical chemistry chemical physics : PCCP, 22(10), 5449-5462 (2020-02-23)
A microfluidic technique is coupled with X-ray fluorescence in order to investigate the origin of the so-called synergy effect observed in liquid-liquid extraction of rare earth elements (REEs) when special combinations of two extractants - one solvating and one ionic
K C Verma et al.
Physical chemistry chemical physics : PCCP, 21(23), 12540-12554 (2019-06-01)
The induction of charge and spin in diluted magnetic semiconductor ZnO is explored for spintronic devices and its wide direct band gap (3.37 eV) and large exciton binding energy (60 meV) exhibit potential in UV photodetectors. We reported the ferromagnetic
Photocatalytic degradation performance of Nd3+ doped V2O5 nanostructures
Navyashree GR, et al.
Materials Research Express, 5(9), 095007-095007 (2018)

Articles

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service