Skip to Content
Merck
All Photos(1)

Key Documents

379115

Sigma-Aldrich

DCC

1.0 M in methylene chloride, for peptide synthesis

Synonym(s):

N,N′-Dicyclohexylcarbodiimide solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H11N=C=NC6H11
CAS Number:
Molecular Weight:
206.33
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Product Name

DCC, 1.0 M in methylene chloride

form

liquid

Quality Level

reaction suitability

reaction type: Coupling Reactions

concentration

1.0 M in methylene chloride

density

1.247 g/mL at 25 °C

application(s)

peptide synthesis

SMILES string

C1CCC(CC1)N=C=NC2CCCCC2

InChI

1S/C13H22N2/c1-3-7-12(8-4-1)14-11-15-13-9-5-2-6-10-13/h12-13H,1-10H2

InChI key

QOSSAOTZNIDXMA-UHFFFAOYSA-N

Gene Information

human ... EPHX2(2053)
mouse ... Ephx2(13850)

Looking for similar products? Visit Product Comparison Guide

Application

DCC (N,N′-Dicyclohexylcarbodiimide) is mainly used to mediate peptide coupling[1] and also to activate carboxylic acids for esterification.[2]

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Carc. 2 - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effects of amounts of additives on peptide coupling mediated by a water?soluble carbodiimide in alcohols.
Nozaki S.
The Journal of Peptide Research, 54(2), 162-167 (1999)
Liposomes containing alkylated methotrexate analogues for phospholipase A 2 mediated tumor targeted drug delivery.
Kaasgaard T, et al.
Chemistry and Physics of Lipids, 157(2), 94-103 (2009)
Denys Pogoryelov et al.
Nature chemical biology, 6(12), 891-899 (2010-10-26)
The microscopic mechanism of coupled c-ring rotation and ion translocation in F(1)F(o)-ATP synthases is unknown. Here we present conclusive evidence supporting the notion that the ability of c-rings to rotate within the F(o) complex derives from the interplay between the
Masashi Toei et al.
The Journal of biological chemistry, 288(36), 25717-25726 (2013-07-31)
N,N-Dicyclohexylcarbodiimide (DCCD) is a classical inhibitor of the F0F1-ATP synthase (F0F1), which covalently binds to the highly conserved carboxylic acid of the proteolipid subunit (c subunit) in F0. Although it is well known that DCCD modification of the c subunit
Marten Staal et al.
Plant physiology, 155(4), 2049-2055 (2011-02-02)
In Arabidopsis (Arabidopsis thaliana; Columbia-0) roots, the so-called zone of cell elongation comprises two clearly different domains: the transition zone, a postmeristematic region (approximately 200-450 μm proximal of the root tip) with a low rate of elongation, and a fast

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service