Skip to Content
Merck
All Photos(2)

Documents

242306

Sigma-Aldrich

Allyl chloroformate

97%, for peptide synthesis

Synonym(s):

2-Propen-1-yl chloroformate, Allyloxycarbonyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCOOCH2CH=CH2
CAS Number:
Molecular Weight:
120.53
Beilstein:
773915
EC Number:
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:
NACRES:
NA.22

product name

Allyl chloroformate, 97%

vapor pressure

3.67 psi ( 20 °C)

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.422 (lit.)

bp

~27 °C/11 mmHg (lit.)

density

1.134 g/mL at 20 °C (lit.)

application(s)

peptide synthesis

storage temp.

−20°C

SMILES string

ClC(=O)OCC=C

InChI

1S/C4H5ClO2/c1-2-3-7-4(5)6/h2H,1,3H2

InChI key

CAEWJEXPFKNBQL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Allyl chloroformate is an organic compound used to synthesize numerous compounds like allyl carbamates and allyl carbonates. Additionally, it can be used for the preparation of α-allyl carbonyls and α, β-unsaturated carbonyls.

Application

Allyl chloroformate is used in the synthesis of:
  • Histones and their analogs.
  • Alloc-TL monomer (N-(allyloxy)carbonylhomocysteine thiolactone) from cyclic thioester (d,l-homocysteine thiolactonehydrochloride).

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

87.8 °F - closed cup

Flash Point(C)

31 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Carlos E Cuevas-Suárez et al.
Journal of the mechanical behavior of biomedical materials, 87, 148-154 (2018-08-03)
Over the past years, significant effort has been dedicated to synthesizing low-shrinking formulations, however, development of dental composites with low volumetric shrinkage continues to be challenging. The purpose of this study was to synthesize a bisphenol allylic derivate (BPhADAC) and
Palladium-catalyzed synthesis of substituted cycloheptane-1,4-diones by an asymmetric ring-expanding allylation (AREA).
Sabrina R Schulz et al.
Angewandte Chemie (International ed. in English), 46(21), 3966-3970 (2007-04-20)
Thiolactone-based polymers for formaldehyde scavenging coatings
Resetco C, et al.
European Polymer Journal, 82, 166-174 (2016)
Allyl Chloroformate
Herrinton PM
Encyclopedia of Reagents for Organic Synthesis, Second Edition (2001)
[Toxic action of allylchloroformate].
E F Oskerko et al.
Gigiena truda i professional'nye zabolevaniia, (5)(5), 51-52 (1984-05-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service