Skip to Content
Merck
All Photos(1)

Key Documents

376078

Sigma-Aldrich

(Isopropenyloxy)trimethylsilane

≥85%

Synonym(s):

2-(Trimethylsiloxy)propene, Acetone enol trimethylsilyl ether, IPOTMS

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C(=CH2)OSi(CH3)3
CAS Number:
Molecular Weight:
130.26
Beilstein:
1849023
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥85%

form

liquid

impurities

10-15% hexamethyldisiloxane

refractive index

n20/D 1.395 (lit.)

density

0.78 g/mL at 25 °C (lit.)

storage temp.

−20°C

SMILES string

CC(=C)O[Si](C)(C)C

InChI

1S/C6H14OSi/c1-6(2)7-8(3,4)5/h1H2,2-5H3

InChI key

UAIFZYSPVVBOPN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(Isopropenyloxy)trimethylsilane is an enol silyl ether. It participates as nucleophile in Lewis acid-promoted C3-nucleophile substitution of tetracyclic bromide. It is reported to participate in the total synthesis of (±)-powelline. It is reported to participate in dityltin bis(triflate) catalyzed Michael addition reaction of enol silyl ethers.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

19.4 °F - closed cup

Flash Point(C)

-7 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Justin Kim et al.
Journal of the American Chemical Society, 133(38), 14940-14943 (2011-08-31)
Concise and enantioselective total syntheses of (+)-naseseazines A and B are described. Our regioselective and directed dimerization of diketopiperazines provides their critical C3-C(sp(2)) linkages, an assembly with plausible biogenetic relevance. We revise the absolute stereochemistry of (+)-naseseazines A and B.
Katherine M Bogle et al.
Organic letters, 12(6), 1252-1254 (2010-02-24)
The total synthesis of (+/-)-powelline (13 linear steps in an overall yield of 6%) and (+/-)-buphanidrine (14 linear steps and a 6% overall yield) and has been achieved using a novel approach to the crinane skeleton. An organocatalytic oxidative coupling
Organotin triflate as practical catalyst for Michael addition of enol silyl ethers.
Sato T, et al.
Tetrahedron, 47(47), 9773-9782 (1991)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service