Skip to Content
Merck
All Photos(1)

Key Documents

415014

Sigma-Aldrich

Vinyloxy-trimethylsilane

97%

Synonym(s):

(Trimethylsiloxy)ethylene, TMSO-ethene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3SiOCH=CH2
CAS Number:
Molecular Weight:
116.23
Beilstein:
1738846
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.389 (lit.)

bp

74 °C (lit.)

density

0.779 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

[H]\C([H])=C(\[H])O[Si](C)(C)C

InChI

1S/C5H12OSi/c1-5-6-7(2,3)4/h5H,1H2,2-4H3

InChI key

HFTNNOZFRQLFQB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

precipitate will not affect use

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-2.2 °F - closed cup

Flash Point(C)

-19 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marcos N Eberlin
Journal of mass spectrometry : JMS, 41(2), 141-156 (2006-02-01)
This article discusses the application of gas-phase ion/molecule reactions for fine structural elucidation in mass spectrometry. This approach is illustrated via a representative collection of class- and functional group-selective reactions, a few of historical relevance as well as by more
Color stability and spectroscopic properties of deoxyvitisins in aqueous solution
Sousa A, et al.
New. J. Chem., 38(2), 539-544 (2014)
Deoxyvitisins: a new set of pyrano-3-deoxyanthocyanidins.
Sousa A, et al.
Tetrahedron Letters, 54(35), 4785-4788 (2013)
A novel synthetic pathway to vitisin B compounds.
Oliveira J, et al.
Tetrahedron Letters, 50(27), 3933-3935 (2009)
Inside the black box-Perspectives on transformations in catalysis.
Fogg DE.
Canadian Journal of Chemistry, 86(10), 931-941 (2008)

Articles

Review SET mechanism in photoredox catalysis for sustainable reactions and rearrangement of oxaziridines.

Review SET mechanism in photoredox catalysis for sustainable reactions and rearrangement of oxaziridines.

Review SET mechanism in photoredox catalysis for sustainable reactions and rearrangement of oxaziridines.

Review SET mechanism in photoredox catalysis for sustainable reactions and rearrangement of oxaziridines.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service