Skip to Content
Merck
All Photos(2)

Key Documents

274585

Sigma-Aldrich

Methyl trimethylsilyl dimethylketene acetal

95%

Synonym(s):

1-Methoxy-2-methyl-1-(trimethylsiloxy)propene, (1-Methoxy-2-methyl-1-propenyloxy)trimethylsilane, MTDA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C=C(OCH3)OSi(CH3)3
CAS Number:
Molecular Weight:
174.31
Beilstein:
1362893
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

liquid

refractive index

n20/D 1.415 (lit.)

bp

35 °C/15 mmHg (lit.)

density

0.858 g/mL at 25 °C (lit.)

SMILES string

CO\C(O[Si](C)(C)C)=C(/C)C

InChI

1S/C8H18O2Si/c1-7(2)8(9-3)10-11(4,5)6/h1-6H3

InChI key

JNOGVQJEBGEKMG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Methyl trimethylsilyl dimethylketene acetal was used:
  • in the synthesis of chiral β-lactams by reacting with (S)-alkylidene(1-arylethyl)amines in the presence of titanium tetrachloride
  • as a catalyst or initiator in the group-transfer polymerization
  • as a versatile reagent in conjugate addition and aldol reactions.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

86.0 °F - closed cup

Flash Point(C)

30 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Tetrahedron Letters, 35, 5239-5239 (1994)
Polymer Journal, 26, 169-169 (1994)
Journal of the American Chemical Society, 116, 4131-4131 (1994)
Journal of Polymer Science Part A: Polymer Chemistry, 45, 2514-2514 (2007)
Asymmetric synthesis of ?-lactams. I. The reaction of dimethylketene silyl acetal with (S)-alkylidene (1-arylethyl) amines promoted by titanium tetrachloride.
Tetrahedron Letters, 21(21), 2077-2080 (1980)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service