Skip to Content
Merck
All Photos(2)

Key Documents

661376

Sigma-Aldrich

2,3-Diaminophenazine

90%

Synonym(s):

2,3-Phenazinediamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H10N4
CAS Number:
Molecular Weight:
210.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

90%

form

solid

mp

>300 °C

SMILES string

Nc1cc2nc3ccccc3nc2cc1N

InChI

1S/C12H10N4/c13-7-5-11-12(6-8(7)14)16-10-4-2-1-3-9(10)15-11/h1-6H,13-14H2

InChI key

VZPGINJWPPHRLS-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Watanabe et al.
Mutation research, 245(1), 15-22 (1990-09-01)
The mutagenicity of o- and m-phenylenediamine (PD) was remarkedly enhanced by oxidation; their major mutagenic oxidation products were 2,3- and 2,7-diaminophenazine, respectively. In order to evaluate the modulation effect of p-PD on the oxidation of m- or o-PD, p-PD and
Patty K-L Fu et al.
Photochemistry and photobiology, 81(1), 89-95 (2004-10-21)
Aromatic amines, such as o-phenylenediamine (OPD), have been used extensively in commercial hair dyes and in the synthesis of agricultural pesticides. Air oxidation of OPD results in the formation of 2,3-diaminophenazine (DAP). Although the mutagenic toxicity of DAP has been
P S Oturai et al.
Metabolism: clinical and experimental, 49(8), 996-1000 (2000-08-23)
The accelerated formation of advanced glycation end-products (AGEs) due to elevated glycemia has repeatedly been reported as a central pathogenic factor in the development of diabetic microvascular complications. The effects of a novel inhibitor of AGE formation, NNC39-0028 (2,3-diaminophenazine), and
R P Doyle et al.
Acta crystallographica. Section C, Crystal structure communications, 57(Pt 1), 104-105 (2001-02-15)
The planar electron-rich heterocyclic diamine 2,3-diaminophenazine (DAP), C12H10N4, is of particular interest to both chemists and biochemists because of its rich organic chemistry and intense luminescence. In this paper, we report the first structure of DAP in its non-protonated form
G Grosa et al.
Xenobiotica; the fate of foreign compounds in biological systems, 34(4), 345-352 (2004-07-23)
1. The results of an in vitro study of the metabolism of benzofuroxan using either cytosolic or microsomal fractions obtained from rat liver are reported. 2. Benzofuroxan was incubated with an appropriate volume of cytosol or microsomal suspension; control incubations

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service