Skip to Content
Merck
All Photos(2)

Key Documents

363170

Sigma-Aldrich

Poly(vinyl alcohol)

Poly(vinyl alcohol)

Mw 13,000-23,000, 87-89% hydrolyzed

Synonym(s):

PVA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-CH2CHOH-]n
CAS Number:
MDL number:
UNSPSC Code:
12352104
NACRES:
NA.23
Pricing and availability is not currently available.

Quality Level

form

crystalline powder
crystals or granules
powder

mol wt

Mw 13,000-23,000

falling ball

3.5-4.5 cP, 4 % in H2O(20 °C)(lit.)

InChI

1S/C2H4O/c1-2-3/h2-3H,1H2

InChI key

IMROMDMJAWUWLK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Polyvinyl alcohol(PVA) is a linear hydrophilicsynthetic polymer with a monoclinic crystal structure. It can be synthesized via the polymerization of vinyl esters orethers, with subsequent saponification or transesterification. Due to itsexcellent physical and chemical properties, PVA has been widely used inadhesives, fibers, paper industry, drug delivery, and tissue engineering.

Application

PVA is used as a biological reagent to test the dispersion of zinc oxide nanoparticles.[1] PVA has been evaluated as an effective protein resistant material.2 It may be used to stabilize gold colloids of diameter 3-8nm.[2]
Poly(vinyl alcohol) can be used as a secondary template in the synthesis of mesoporous zeolites.

It can be used to prepare a hydrogel template to produce homogeneous microparticles for sustained drug release. PVA is used as a stabilizer in micro-particle preparation due to its high tensile strength and flexibility.

Features and Benefits

  • Good mechanical and chemical stability
  • Non-toxic
  • Tunable hydrophilicity
  • Resistance to solvents Capable of forming films

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

No data available

Flash Point(C)

No data available

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gold colloidal nanoparticles sized to be suitable precursors for heterogeneous catalysts
Porta F and Prati L
Metal Nanoclusters in Catalysis and Materials Science: The Issue of Size Control, 355-355 (2008)
Zinc oxide nanoparticles as selective killers of proliferating cells.
Taccola L, et al.
International journal of nanomedicine, 6, 1129-1140 (2010)
E Fortunati et al.
Carbohydrate polymers, 97(2), 825-836 (2013-08-06)
PVA bio-nanocomposites reinforced with cellulose nanocrystals (CNC) extracted from commercial microcrystalline cellulose (MCC) and from two types of natural fibres, Phormium tenax and Flax of the Belinka variety, were produced by solvent casting in water. Morphological, thermal, mechanical and transparency
Xiao-Zhu Sun et al.
Carbohydrate polymers, 94(1), 147-153 (2013-04-03)
Polyvinyl alcohol (PVA) nanofibers loaded with curcumin or its β-cyclodextrin (CD) inclusion complex were successfully prepared using an electrospinning process. The influence of curcumin or CD-curcumin complex content on fiber formation and quality was investigated. X-ray diffraction and differential scanning
Khoon S Lim et al.
Biomaterials, 34(29), 7097-7105 (2013-06-27)
Development of tissue engineering solutions for biomedical applications has driven the need for integration of biological signals into synthetic materials. Approaches to achieve this typically require chemical modification of the biological molecules. Examples include chemical grafting of synthetic polymers onto

Questions

1–3 of 3 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. Do you have any data indicating an expected range of particle size for PVA, Poly(vinyl alcohol), products 363170 and 363138 (CAS 9002-89-5)?

    1 answer
    1. The particle size is an internal specification, and there are no external specifications beyond the internal “S” grade. Internally, the focus is on ensuring there are no excessive quantities of fines (<100 Mesh) or excessive large particles (>14 Mesh). These internal requirements do not vary between the two products listed.

      Helpful?

Reviews

Active Filters

  1. 1 Ratings-only review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service