Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Poly(vinyl alcohol)
Synonym(s):
Poly(vinyl alcohol)
Linear Formula:
[-CH
2
CHOH-]
n
CAS Number:
9002-89-5
Products
Product Category
polymers (8)
binders (7)
coatings (7)
stabilizers (7)
thickeners (7)
detergents (1)
Brand
SAFC (7)
Sigma-Aldrich (14)
Supelco (1)
Feature
Physical Form
Product Line
Agency
Quality Segment
Application
Polymer Composition
Polymer Type
Technique
Filter Products
Showing 1-20 of 24
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
341584
Poly(vinyl alcohol),
M
w
89,000-98,000, 99+% hydrolyzed
Expand
View Pricing
360627
Poly(vinyl alcohol),
M
w
9,000-10,000, 80% hydrolyzed
Expand
View Pricing
363065
Poly(vinyl alcohol),
M
w
146,000-186,000, 99+% hydrolyzed
Expand
View Pricing
363170
Poly(vinyl alcohol),
M
w
13,000-23,000, 87-89% hydrolyzed
Expand
View Pricing
363138
Poly(vinyl alcohol),
M
w
31,000-50,000, 98-99% hydrolyzed
Expand
View Pricing
363146
Poly(vinyl alcohol),
M
w
85,000-124,000, 99+% hydrolyzed
Expand
View Pricing
P1763
Poly(vinyl alcohol),
Fully hydrolyzed
Expand
View Pricing
1.41351
Poly(vinyl alcohol),
8-88, EMPROVE
®
ESSENTIAL, Ph. Eur., USP, JPE
Expand
View Pricing
P8136
Poly(vinyl alcohol),
87-90% hydrolyzed, average mol wt 30,000-70,000
Expand
View Pricing
363073
Poly(vinyl alcohol),
average M
w
31,000-50,000, 87-89% hydrolyzed
Expand
View Pricing
363081
Poly(vinyl alcohol),
average M
w
85,000-124,000, 87-89% hydrolyzed
Expand
View Pricing
363103
Poly(vinyl alcohol),
average M
w
146,000-186,000, 87-89% hydrolyzed
Expand
View Pricing
348406
Poly(vinyl alcohol),
average M
w
13,000-23,000, 98% hydrolyzed
Expand
View Pricing
1.41350
Poly(vinyl alcohol),
4-88, EMPROVE
®
ESSENTIAL, Ph. Eur., ChP, JPE, USP
Expand
View Pricing
563900
Poly(vinyl alcohol),
average M
w
130,000, 99+% hydrolyzed
Expand
View Pricing
1.41354
Poly(vinyl alcohol),
5-88, EMPROVE
®
ESSENTIAL, Ph. Eur., USP, JPE
Expand
View Pricing
1.41353
Poly(vinyl alcohol),
40-88, EMPROVE
®
ESSENTIAL, Ph. Eur., ChP, USP, JPE
Expand
View Pricing
1.14266
Poly(vinyl alcohol),
protective colloid for argentometric titration
Expand
View Pricing
1.41356
Poly(vinyl alcohol),
28-99, EMPROVE
®
ESSENTIAL, Ph. Eur., JPE
Expand
View Pricing
1548065
Poly(vinyl alcohol),
United States Pharmacopeia (USP) Reference Standard
Expand
View Pricing
Page 1 of 2
Page 1 of 2