Skip to Content
Merck
All Photos(1)

Documents

81381

Sigma-Aldrich

Mowiol® 4-88

Mw ~31,000

Synonym(s):

Poly(vinyl alcohol)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-CH2CHOH-]n
CAS Number:
MDL number:
UNSPSC Code:
12352104
NACRES:
NA.23

form

crystals

Quality Level

mol wt

Mw ~31,000

extent of labeling

86.7-88.7 mol% hydrolysis
~630 polymerization

ign. residue

≤0.5%

viscosity

3.5-4.5 mPa.s, 4 % in H2O(20 °C)

InChI

1S/C2H4O/c1-2-3/h2-3H,1H2

InChI key

IMROMDMJAWUWLK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Mowiol® is a water soluble hydrocolloid mucoadhesive based on poly (vinyl alcohol). Mowiol® at pH 8.5 has a couple of advantages over other mounting media:
  • its a liquid when the sample is mounted and solidifies after several hours, forming a permanent bond between the coverslip and slide
  • fluorescence is stable when held in dark and at 4oC.

Application

Mowiol® is used as an embedding medium for preserving stained cell dishes in the study of siRNA array production of HeLa cells. Mowiol® is also reported to be used as a mounting medium in:
  • design of TSAR and its applications
  • detection of the secreted and cytoplasmic fractions of IpaB, IpaC and IpaD by lysozyme permeabilization
  • fluorescence recovery after photobleaching (FRAP).

Mowiol® may be used as a coating for sensors based on piezoelectric quartz crystals. Another probable use of Mowiol® is during the study of super fluorescence imaging of biocompatible carbon dots.

Legal Information

Mowiol is a registered trademark of Kuraray Europe GmbH

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

No data available

Flash Point(C)

No data available

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaofang Li et al.
Scientific reports, 7(1), 16495-16495 (2017-12-01)
Heterogeneity in terms of tumor characteristics, prognosis, and survival among cancer patients is an unsolved issue. Here, we systematically analyzed the aberrant expression patterns of cervical cancer using RNA-Seq data from The Cancer Genome Atlas (TCGA). We incorporated gene profiling
Avgi Tsolou et al.
Life sciences, 232, 116562-116562 (2019-06-16)
Lung cancer is one of the main causes of cancer-related deaths worldwide and radiotherapy is a major treatment of choice. However, radioresistance is a main reason for radiotherapy failure or tumor relapse. Here, we investigated possible mechanisms associated with cancer
Super-resolution fluorescence imaging of biocompatible carbon dots.
Lemenager G, et al.
Nanoscale, 6, 8614-8623 (2014)
Cedric S Tremblay et al.
Nature communications, 9(1), 3535-3535 (2018-09-01)
Pre-leukemic stem cells (pre-LSCs) give rise to leukemic stem cells through acquisition of additional gene mutations and are an important source of relapse following chemotherapy. We postulated that cell-cycle kinetics of pre-LSCs may be an important determinant of clonal evolution
A Kondratskyi et al.
Cell death & disease, 5, e1193-e1193 (2014-04-26)
The growing number of studies suggested that inhibition of autophagy enhances the efficacy of Akt kinase inhibitors in cancer therapy. Here, we provide evidence that ML-9, a widely used inhibitor of Akt kinase, myosin light-chain kinase (MLCK) and stromal interaction

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service