Skip to Content
Merck
All Photos(1)

Key Documents

35840

Sigma-Aldrich

Methyl dichloroacetate

≥99%

Synonym(s):

Dichloroacetic acid methyl ester, NSC 2882

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2CHCOOCH3
CAS Number:
Molecular Weight:
142.97
Beilstein:
1747542
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99%

form

liquid

refractive index

n20/D 1.442 (lit.)

bp

143 °C (lit.)

mp

−52 °C (lit.)

density

1.381 g/mL at 25 °C (lit.)

SMILES string

COC(=O)C(Cl)Cl

InChI

1S/C3H4Cl2O2/c1-7-3(6)2(4)5/h2H,1H3

InChI key

HKMLRUAPIDAGIE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl dichloroacetate undergoes base-catalyzed condensation with aliphatic α-haloaldehydes. Chlorination of methyl dichloroacetate by using trifluoromethanesulfonyl chloride has been reported.

Application

Methyl dichloroacetate may be used:
  • in the diastereoselctive synthesis of bicyclic chlorocyclopropane
  • as difunctional initiator during atom transfer radical polymerization of methyl or n-butyl acrylate
  • as reagent during CrCl2-induced olefination of aldehydes
Reactant involved in:
  • Nucleophilic substitution reactions with nitro(pentafluorosulfanyl)benzenes
  • Azide-alkyl halide cycloaddition and atom transfer radical addition
  • Chemical modification of STn antigen for enhancement of synthetic carbohydrate vaccines
  • Three component reaction of arynes with cyclic ethers and active methines
  • Catalytic radical haloalkylation for synthesis of neodysidenin
  • Addition reactions for syntehsis of enantiopure 3,4-diamino esters or amides

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

147.2 °F - closed cup

Flash Point(C)

64 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Diastereoselective cyclopropanation of cyclic enones with methyl dichloroacetate anion.
Escribano A, et al.
Tetrahedron, 57(46), 9423-9427 (2001)
A highly stereospecific synthesis of (E)-a, ?-unsaturated esters.
Barma DK, et al.
Tetrahedron Letters, 45(30), 5917-5920 (2004)
Trifluoromethanesulfonyl chloride, a mild chlorinating agent.
Hosein Hakimelahi G and Just G.
Tetrahedron Letters, 20(38), 3643-3644 (1979)
Synthesis of telechelic polyacrylates with unsaturated end-groups.
Bielawski CW, et al.
Polymer, 44(13), 3721-3726 (2003)
The Base-catalyzed Condensation of a-Haloaldehydes with Dichloroacetates.
Takeda A, et al.
Bulletin of the Chemical Society of Japan, 45(4), 1217-1220 (1972)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service