Skip to Content
Merck
All Photos(1)

Key Documents

35820

Sigma-Aldrich

Ethyl dichloroacetate

≥99.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2CHCOOC2H5
CAS Number:
Molecular Weight:
157.00
Beilstein:
773995
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥99.0% (GC)

refractive index

n20/D 1.438

bp

54-55 °C/11 mmHg (lit.)

density

1.28 g/mL at 20 °C (lit.)

SMILES string

CCOC(=O)C(Cl)Cl

InChI

1S/C4H6Cl2O2/c1-2-8-4(7)3(5)6/h3H,2H2,1H3

InChI key

IWYBVQLPTCMVFO-UHFFFAOYSA-N

General description

Ethyl dichloroacetate is an ester and its base catalyzed hydrolysis has been reported.

Application

Ethyl dichloroacetate may be used in the synthesis of α-halo-carboxylic acid ester.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

143.6 °F - closed cup

Flash Point(C)

62 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

General Base Catalysis of Ester Hydrolysis1.
Jencks WP and Carriuolo J.
Journal of the American Chemical Society, 83(7), 1743-1750 (1961)
Reaction of B-alkyl-9-borabicyclo [3.3. 1] nonanes with ethyl mono-and dihaloacetates under the influence of potassium tert-butoxide. A convenient procedure for the conversion of olefins into esters or. alpha.-halo esters via hydroboration.
Brown HC and Milorad M. Rogic.
Journal of the American Chemical Society, 91(8), 2146-2147 (1969)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service