Skip to Content
Merck
All Photos(2)

Documents

403067

Sigma-Aldrich

Sodium methoxide solution

ACS reagent, 0.5 M CH3ONa in methanol (0.5N)

Synonym(s):

Sodium methylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3ONa
CAS Number:
Molecular Weight:
54.02
Beilstein:
3592982
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent

Quality Level

form

liquid

concentration

0.5 M CH3ONa in methanol (0.5N)

color

, clarity of solution to pass

bp

64.6 °C

density

0.809 g/mL at 25 °C

SMILES string

[Na+].C[O-]

InChI

1S/CH3O.Na/c1-2;/h1H3;/q-1;+1

InChI key

WQDUMFSSJAZKTM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sodium methoxide solution is composed of 0.5M sodium methoxide in 0.5N methanol. It is a widely used organic base in organic synthesis.

Application

Sodium methoxide solution may be used in the following processes:
  • As a solvent in the synthesis of disulfides from corresponding thiols.
  • To extract fatty acid methyl esters by transesterification process.
  • Synthesis of (1R,2R,3R,7aS)-1,2-dibenzyloxy-3-tert-butyldiphenylsilyloxymethylpyrrolizidin-5-one.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Polyhydroxylated pyrrolizidines. Part I: Short and highly stereocontrolled syntheses of hyacinthacines.
Izquierdo I, et al.
Tetrahedron Asymmetry, 12(17), 2481-2487 (2001)
Biosynthetic origin of conjugated double bonds: production of fatty acid components of high-value drying oils in transgenic soybean embryos.
Cahoon EB, et al.
Proceedings of the National Academy of Sciences of the USA, 96(22), 12935-12940 (1999)
Electrochemical oxidation of thiols to disulfides.
Leite SLS, et al.
Synthetic Communications, 20(3), 393-397 (1990)
Gema Flores et al.
Food chemistry, 194, 1260-1265 (2015-10-17)
Black currant seeds are obtained as a residue during juice production. Black currant seed oil contains high amounts of nutritionally desirable constituents such as γ-linolenic acid (GLA), α-linolenic acid (ALA) and stearidonic acid (SA), as well as certain phenolic acids
Holger Pletsch et al.
Nanoscale, 7(5), 1977-1983 (2014-12-24)
The design of networks from polymers and noble metal nanoparticles requires thorough control over topological polymer-particle arrangements. This study explores the interaction between a linear polydentate poly(propylene sulfide) (PPrS) ligand and silver nanoparticles (AgNPs) with an aim to study its

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service