Skip to Content
Merck
All Photos(1)

Key Documents

338338

Sigma-Aldrich

Fmoc-Phe-OH

98%, for peptide synthesis

Synonym(s):

N-(9-Fluorenylmethoxycarbonyl)-L-phenylalanine, Fmoc-L-phenylalanine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H21NO4
CAS Number:
Molecular Weight:
387.43
Beilstein:
3597808
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26

Product Name

Fmoc-Phe-OH, 98%

Assay

98%

optical activity

[α]20/D −37°, c = 1 in DMF

reaction suitability

reaction type: C-H Activation
reaction type: Fmoc solid-phase peptide synthesis
reagent type: ligand
reaction type: Peptide Synthesis

mp

180-187 °C (lit.)

application(s)

peptide synthesis

functional group

Fmoc
amine
carboxylic acid

storage temp.

2-8°C

SMILES string

OC(=O)[C@H](Cc1ccccc1)NC(=O)OCC2c3ccccc3-c4ccccc24

InChI

1S/C24H21NO4/c26-23(27)22(14-16-8-2-1-3-9-16)25-24(28)29-15-21-19-12-6-4-10-17(19)18-11-5-7-13-20(18)21/h1-13,21-22H,14-15H2,(H,25,28)(H,26,27)/t22-/m0/s1

InChI key

SJVFAHZPLIXNDH-QFIPXVFZSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Roman Schroeder et al.
Analytical biochemistry, 588, 113488-113488 (2019-10-28)
Cathepsin G (CatG) is responsible for several distinct immune processes of adaptive and innate immunity depending on extra- or intracellular occurrence of CatG. Recently, we established a method to detect CatG activity at the cell surface of natural killer cells
R Ashton Lavoie et al.
Biotechnology and bioengineering, 117(2), 438-452 (2019-10-28)
The clearance of host cell proteins (HCPs) is of crucial importance in biomanufacturing, given their diversity in composition, structure, abundance, and occasional structural homology with the product. The current approach to HCP clearance in the manufacturing of monoclonal antibodies (mAbs)
Alice P McCloskey et al.
Journal of peptide science : an official publication of the European Peptide Society, 23(2), 131-140 (2017-01-10)
Biomaterial-related infections have a significant impact on society and are a major contributor to the growing threat of antimicrobial resistance. Current licensed antibiotic classes struggle to breakdown or penetrate the exopolysaccharide biofilm barrier, resulting in sub-therapeutic concentrations of antibiotic at
Srinivasa R Tala et al.
ACS chemical neuroscience, 9(5), 1001-1013 (2017-12-20)
The melanocortin system is involved in the regulation of complex physiological functions, including energy and weight homeostasis, feeding behavior, inflammation, sexual function, pigmentation, and exocrine gland function. The five melanocortin receptors that belong to the superfamily of G protein-coupled receptors
Chiao-Ting Yen et al.
European journal of medicinal chemistry, 44(5), 1933-1940 (2008-12-27)
Twenty-four new dipeptide analogs (1-24) of aurantiamide acetate were designed, synthesized, and assayed for effects on superoxide anion generation and elastase release by human neutrophils in response to fMLP/CB. Among them, seven N-(fluorenyl-9-methoxycarbonyl) (Fmoc)-dipeptides (6, 9, 12, 14, 17, 18

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service