All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C17H15NO4
CAS Number:
Molecular Weight:
297.31
Beilstein:
2163967
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26
Recommended Products
Assay
≥98.0% (T)
form
powder
reaction suitability
reaction type: Fmoc solid-phase peptide synthesis
mp
174-175 °C (lit.)
174-178 °C
application(s)
peptide synthesis
functional group
Fmoc
storage temp.
2-8°C
SMILES string
OC(=O)CNC(=O)OCC1c2ccccc2-c3ccccc13
InChI
1S/C17H15NO4/c19-16(20)9-18-17(21)22-10-15-13-7-3-1-5-11(13)12-6-2-4-8-14(12)15/h1-8,15H,9-10H2,(H,18,21)(H,19,20)
InChI key
NDKDFTQNXLHCGO-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
- Efficient Fmoc-Protected Amino Ester Hydrolysis Using Green Calcium (II) Iodide as a Protective Agent: This study describes the utilization of environmentally friendly calcium iodide in the hydrolysis of Fmoc-protected amino esters, enhancing reaction efficiency and sustainability (R Binette, M Desgagné, C Theaud, PL Boudreault - Molecules, 2022). Link to the article.
- α/β-Chimera peptide synthesis with cyclic β-sugar amino acids: the efficient coupling protocol: This research provides an advanced synthesis method for α/β-chimera peptides using cyclic β-sugar amino acids, demonstrating significant implications for peptide design in medicinal chemistry (A Nagy, V Goldschmidt Gőz, I Pintér, V Farkas - Amino acids, 2019). Link to the article.
- MS, CD, and FT-IR characterization of five newly synthesized histidine-containing Ala-and Gly-based peptides: This paper presents detailed characterization of novel histidine-containing peptides, highlighting techniques that could be pivotal for peptide-based drug discovery (M Murariu, L Ion, CI Ciobanu, BA Petre - Rev. Roum Chem., 2017). Link to the article.
- Efficient method for the concentration determination of fmoc groups incorporated in the core-shell materials by Fmoc–glycine: This article elaborates on an efficient method for determining the concentration of fmoc groups in core-shell materials, critical for the design of advanced functional materials (E Szczepańska, B Grobelna, J Ryl, A Kulpa - Molecules, 2020). Link to the article.
- Circular aqueous fmoc/t‐bu solid‐phase peptide synthesis: This study explores a novel approach in solid-phase peptide synthesis, utilizing circular aqueous techniques that may offer greener and more efficient methodologies for peptide synthesis (J Pawlas, JH Rasmussen - ChemSusChem, 2021). Link to the article.
Other Notes
Fmoc-glycine coupling of saccharide β-glycosylamines for the fractionation of oligosaccharides and formation of neoglycoconjugates.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Jing Fu et al.
Ecotoxicology and environmental safety, 189, 110039-110039 (2019-12-13)
The omnipresence of antimicrobial triclosan (TCS) and by-products in aquatic environments is a threat to aquatic organisms. Traditionally, the adverse effects of TCS and its by-products have been evaluated by examining the phenotypic output relevant to predicting acute toxicity rather
Jing Fu et al.
Journal of hazardous materials, 368, 186-196 (2019-01-25)
Methyl-triclosan (MTCS), as a biodegradation product from antibacterial triclosan (TCS), has been detected in water catchments, and it has also been verified to accumulate in biota due to its hydrophobicity. There is a lack, however, of toxicity studies on MTCS
L Noronha-Blob et al.
Gastroenterology, 104(4), 1021-1029 (1993-04-01)
The efficacy of the leukocyte recruitment inhibitor, N-[9H-2,7-dimethylfluoren-9-ylmethoxy)carbonyl]-L-leucine (NPC 15669) was compared with drugs used to treat inflammatory bowel diseases in a rat model, acetic acid-induced colitis. Colonic damage assessed by visual inspection, histological quantitation of tissue injury, vascular permeability
Mario Fernández-Fernández et al.
Journal of mass spectrometry : JMS, 51(10), 980-987 (2016-07-09)
We have developed a novel, rapid and easy calculation procedure for Mass Isotopomer Distribution Analysis based on multiple linear regression which allows the simultaneous calculation of the precursor pool enrichment and the fraction of newly synthesized labelled proteins (fractional synthesis)
G Arsequell et al.
Analytical biochemistry, 216(1), 165-170 (1994-01-01)
We have devised a new saccharide derivatization scheme to provide not only the temporary attachment of a chromophore for detecting and facilitating the chromatographic separation of carbohydrates, but also the intermediates for further derivatization to produce neoglycoconjugates. Several neutral unprotected
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service