Skip to Content
Merck
All Photos(1)

Key Documents

15480

Sigma-Aldrich

Boc-Phe-OH

≥99.0% (T)

Synonym(s):

N-(tert-Butoxycarbonyl)-L-phenylalanine, Boc-L-phenylalanine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CH(COOH)NHCOOC(CH3)3
CAS Number:
Molecular Weight:
265.30
Beilstein:
2219729
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99.0% (T)

form

solid

optical activity

[α]20/D +25±1°, c = 1% in ethanol

reaction suitability

reaction type: Boc solid-phase peptide synthesis
reaction type: C-H Activation
reagent type: ligand
reaction type: Peptide Synthesis

mp

85-87 °C (lit.)

application(s)

peptide synthesis

functional group

amine
carboxylic acid

SMILES string

CC(C)(C)OC(=O)N[C@@H](Cc1ccccc1)C(O)=O

InChI

1S/C14H19NO4/c1-14(2,3)19-13(18)15-11(12(16)17)9-10-7-5-4-6-8-10/h4-8,11H,9H2,1-3H3,(H,15,18)(H,16,17)/t11-/m0/s1

InChI key

ZYJPUMXJBDHSIF-NSHDSACASA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yong Chen et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 99, 103-113 (2015-12-22)
The objective was to investigate the topical iontophoretic delivery of a series of amino acid ester prodrugs of aciclovir (ACV-X, where ACV=aciclovir and X=Arg, Gly, Ile, Phe, Trp and Val) as a means to enhance cutaneous delivery of ACV. The
Matthew J Crawford et al.
Journal of nucleic acids, 2011, 652702-652702 (2011-07-22)
Conformationally preorganized peptide nucleic acids (PNAs) have been synthesized through backbone modifications at the γ-position, where R = alanine, valine, isoleucine, and phenylalanine side chains. The effects of these side-chains on the conformations and hybridization properties of PNAs were determined
Jie Chen et al.
Archives of pharmacal research, 37(1), 142-148 (2013-11-23)
Novel thermo- and pH-dual responsive amphiphilic copolymers were synthesized based on hyperbranched polyethylenimine (PEI) by grafting L-phenylalanine. The phenylalanine-modified PEI exhibited lower cytotoxicity than commercial PEI. These copolymers showed the phenomena of phase transitions in response to pH and temperature.
Yan Yan et al.
Drug design, development and therapy, 13, 681-694 (2019-03-13)
Budesonide (Bud) is a nonhalogenated glucocorticoid with high anti-inflammatory potency and low systemic side effects. However, the poor water solubility of Bud affects its dissolution and release behavior, thus influencing its anti-inflammatory effect. This study was aimed at synthesizing and
Xiaoxiong Han et al.
Journal of biomaterials science. Polymer edition, 27(7), 626-642 (2016-02-26)
Cabazitaxel (CTX) is a second-generation semisynthetic taxane that demonstrates antitumor activity superior to docetaxel. However, the low aqueous solubility of CTX has hampered its use as a therapeutic agent. In this work, CTX-loaded N-t-butoxycarbonyl-L-phenylalanine end-capped monomethyl poly (ethylene glycol)-block-poly (D,L-lactide)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service