277932
Thallium
granular, ≤6 mm diameter, 99.9% trace metals basis
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
Quality Level
Assay
99.9% trace metals basis
form
granular
resistivity
18 μΩ-cm, 20°C
diameter
≤6 mm
bp
1457 °C (lit.)
mp
303 °C (lit.)
SMILES string
[Tl]
InChI
1S/Tl
InChI key
BKVIYDNLLOSFOA-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Chronic 4 - STOT RE 2
Storage Class Code
6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Environmental science & technology, 46(20), 11086-11094 (2012-09-21)
Thallium (Tl) is emerging as a metal of concern in countries such as China due to its release during the natural weathering of Tl-bearing ore deposits and mining activities. Despite the high toxicity of Tl, few studies have examined the
Inorganic chemistry, 51(23), 13050-13059 (2012-11-20)
Reactions of the extremely bulky amido alkali metal complexes, [KL'(η(6)-toluene)], or in situ generated [LiL'] or [LiL″] {L'/ L″ = N(Ar*)(SiR(3)), where Ar* = C(6)H(2){C(H)Ph(2)}(2)Me-2,6,4 and R = Me (L') or Ph (L″)} with group 13 metal(I) halides have yielded
Physics in medicine and biology, 57(24), 8377-8391 (2012-12-04)
75 RG2 glioma-carrying Fischer rats were treated by photon activation therapy (PAT) with monochromatic synchrotron radiation and stable thallium. Three groups were treated with thallium in combination with radiation at different energy; immediately below and above the thallium K-edge, and
Journal of physics. Condensed matter : an Institute of Physics journal, 25(7), 075703-075703 (2013-01-25)
The effect of selenium substitution by sulfur in the Tl(1-y)Fe(2-z)Se(2) antiferromagnet was studied by x-ray and electron diffraction, magnetization and transport measurements. Tl(0.8)Fe(1.5)(Se(1-x)S(x))(2) (nominal composition) solid solution was synthesized in the full x range (0 ≤ x(S) ≤ 1) using
Lab on a chip, 12(21), 4249-4256 (2012-08-21)
Reproducing the physiological environment of blood vessels for the in vitro investigation of endothelial cell functions is very challenging. Here, we describe a vascular-like structure based on a three-dimensional (3D) gelatin chip with good compatibility and permeability which is also
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service