Skip to Content
Merck
All Photos(2)

Key Documents

208191

Sigma-Aldrich

Thallium(I) sulfate

≥99.9% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Tl2SO4
CAS Number:
Molecular Weight:
504.83
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

≥99.9% trace metals basis

form

powder and chunks

impurities

≤1000.0 ppm Trace Metal Analysis

mp

632 °C (lit.)

density

6.77 g/mL at 25 °C (lit.)

SMILES string

[Tl+].[Tl+].[O-]S([O-])(=O)=O

InChI

1S/H2O4S.2Tl/c1-5(2,3)4;;/h(H2,1,2,3,4);;/q;2*+1/p-2

InChI key

YTQVHRVITVLIRD-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Acute Tox. 3 Dermal - Aquatic Acute 1 - Aquatic Chronic 2 - Skin Irrit. 2 - STOT RE 1 Oral

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Altered testicular enzyme patterns in rats after long-term exposure to thallium sulphate.
C Gregotti et al.
Journal de toxicologie clinique et experimentale, 5(4), 265-271 (1985-07-01)
D B Peele et al.
Neurobehavioral toxicology and teratology, 8(3), 273-277 (1986-05-01)
Flavor aversions induced by intraperitoneal (IP) and oral (PO) administration of thallium sulfate were compared in a repeated trial, two-bottle preference test. Male Long-Evans rats (N = 6/group) were given 30-min access to a 0.1% saccharin solution followed 20-min later
Ritu Tyagi et al.
Journal of applied toxicology : JAT, 31(7), 663-670 (2011-01-11)
Thallium (Tl) is a toxic heavy metal and its exposure to the human body causes physiological and biochemical changes due to its interference with potassium-dependent biological reactions. A high-resolution (1)H NMR spectroscopy based metabonomic approach has been applied for investigating
D Appenroth et al.
Toxicology, 96(3), 203-215 (1995-02-27)
Until now the effect of thallium (Tl) on renal function has not been investigated systematically. Therefore, the dose (5, 10, 15, 20 mg Tl2SO4/kg body wt., intraperitoneally) and time-dependence of renal damage was investigated in diuresis experiments on conscious rats.
C Fleck et al.
Toxicology, 106(1-3), 229-236 (1996-01-08)
The effect of Tl2SO4 (Tl, 2 mg/100 g b.wt.) on renal amino acid excretion and plasma amino acid composition was investigated in 10- and 55-day-old rats. Tl decreased glomerular filtration rate only in adult rats. On the other hand, the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service