Skip to Content
Merck
All Photos(3)

Key Documents

329541

Sigma-Aldrich

Dicumyl peroxide

98%

Synonym(s):

Bis(1-methyl-1-phenylethyl) peroxide, Bis(α,α-dimethylbenzyl) peroxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[C6H5C(CH3)2]2O2
CAS Number:
Molecular Weight:
270.37
Beilstein:
2056090
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

vapor density

9.3 (vs air)

vapor pressure

15.4 mmHg ( 38 °C)

Assay

98%

form

solid

reaction suitability

reagent type: oxidant

mp

39-41 °C (lit.)

density

1.56 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(C)(OOC(C)(C)c1ccccc1)c2ccccc2

InChI

1S/C18H22O2/c1-17(2,15-11-7-5-8-12-15)19-20-18(3,4)16-13-9-6-10-14-16/h5-14H,1-4H3

InChI key

XMNIXWIUMCBBBL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Dicumyl peroxide (DCP) is used:
  • in vulcanization of rubber
  • as a crosslinking agent in the synthesis of polylactic acid composite fibers
  • in the preparation of polyethylene composites
  • in the synthesis of polyamide 112/ethylene vinyl acetate copolymer blends.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Org. Perox. F - Repr. 1B - Skin Irrit. 2

Storage Class Code

5.2 - Organic peroxides and self-reacting hazardous materials

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Lu Lu et al.
Scientific reports, 8(1), 14233-14233 (2018-09-27)
Polymers with reversible elongation upon cooling (EUC) and contraction upon heating (CUH) enabled applications in actuators, fasteners, dampers, grippers, swimmers, sealants, etc. With the current working temperature being limited to mainly above zero Celsius, applications for subzero Celsius environments are
Milan Parchovianský et al.
Materials (Basel, Switzerland), 13(3) (2020-02-07)
In this work, the influence of different cleaning procedures on adhesion of composite coatings containing passive ceramic and commercial glasses was investigated. Two compositions (C2c, D2-PP) of double-layer polymer-derived ceramic (PDC) coating systems, composed from bond coat and a top
Chao Wu et al.
Polymers, 11(4) (2019-04-24)
Silicone rubber composites filled with nano-silica are currently widely used as high voltage insulating materials in power transmission and substation systems. We present a systematic study on the dielectric and mechanical performance of silicone rubber filled with surface modified and
Effect of dynamic cross-linking on phase morphology and dynamic mechanical properties of polyamide 12/ethylene vinyl acetate copolymer blends
Bondan F, et al.
Polym. Bull. null
The effect of maleic anhydride grafting efficiency on the flexural properties of polyethylene composites
Gunning MA, et al.
Journal of Applied Polymer Science, 124(6), 4799-4808 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service