Skip to Content
Merck
All Photos(3)

Documents

287512

Sigma-Aldrich

3-Aminophenylboronic acid monohydrate

98%

Synonym(s):

3-Aminobenzeneboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H4B(OH)2 · H2O
CAS Number:
Molecular Weight:
154.96
Beilstein:
2936342
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

solid

mp

93-96 °C (lit.)

SMILES string

[H]O[H].Nc1cccc(c1)B(O)O

InChI

1S/C6H8BNO2.H2O/c8-6-3-1-2-5(4-6)7(9)10;/h1-4,9-10H,8H2;1H2

InChI key

XAEOVQODHLLNKX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reagent used for
  • Suzuki-Miyaura cross-coupling

Reagent used for Preparation of
  • Gram-positive antivirulence drugs and inhibitors of Streptococcus agalactiae Stk1
  • Regioisomer of Zaleplon (a sedative)
  • Amphiphilic random glycopolymer, which self-assemble to form nanoparticles, with potential as a glucose-sensitive matrix
  • Chemomechanical polymer that expands and contracts in blood plasma with high glucose selectivity

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jing Zhang et al.
Journal of separation science, 40(5), 1107-1114 (2017-01-04)
Novel 3-aminophenylboronic acid functionalized poly(glycidyl methacrylate-co-ethylene dimethacrylate) microspheres were prepared for the solid-phase extraction of glycopeptides/glycoproteins. The adsorption efficiency, maximum adsorption capacity, and specific recognition of the microspheres to glycoprotein were investigated. The results indicated excellent adsorption of glycoproteins by
A chemomechanical polymer that functions in blood plasma with high glucose selectivity.
George K Samoei et al.
Angewandte Chemie (International ed. in English), 45(32), 5319-5322 (2006-08-24)
Sungjin Jung et al.
Advanced healthcare materials, 9(22), e2001633-e2001633 (2020-10-20)
Nanoparticle (NP)-based drug delivery systems or nanomedicines have broadened the horizon of translational research for decades. Conventional bulk mixing synthesis methods have impeded successful clinical translations of nanomedicines due to the limited ability of the controlled, scalable production with high
Gizem Ertürk Bergdahl et al.
Applied biochemistry and biotechnology, 189(2), 374-383 (2019-04-26)
A capacitive sensor was developed to analyze the presence and enzymatic activity of a model protease from standard solutions by following the degradation of the substrate in real time. The enzyme was chosen based on its specific digestion of the
Jiangying Zhu et al.
Materials science & engineering. C, Materials for biological applications, 117, 111273-111273 (2020-09-14)
In this work, poly(ethylene glycol)-b-poly[3-acrylamidophenylboronic acid-co-styrene] (PEG-b-P(PBA-co-St) has been firstly synthesized for loading of insulin to form insulin-loaded micelles. Insulin-loaded micelles (ILM) and epidermal growth factor (EGF) are further embedded into the composite hydrogels that can be rapidly gelled by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service