Skip to Content
Merck
All Photos(3)

Documents

431966

Sigma-Aldrich

4-Formylphenylboronic acid

≥95.0%

Synonym(s):

4-(Dihydroxyboryl)benzaldehyde, 4-Boronobenzaldehyde, 4-Formylbenzeneboronic acid, p-Formylbenzeneboronic acid, p-Formylphenylboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCOC6H4B(OH)2
CAS Number:
Molecular Weight:
149.94
Beilstein:
3030770
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95.0%

mp

237-242 °C (lit.)

SMILES string

OB(O)c1ccc(C=O)cc1

InChI

1S/C7H7BO3/c9-5-6-1-3-7(4-2-6)8(10)11/h1-5,10-11H

InChI key

VXWBQOJISHAKKM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Formylphenylboronic acid is a substrate for Suzuki cross-coupling reactions and it can be used as a reagent for:
  • Palladium-catalyzed Suzuki-Miyaura cross-coupling in water.
  • Copper-mediated ligandless aerobic fluoroalkylation of arylboronic acids with fluoroalkyl iodides.
  • Ligand-free copper-catalyzed coupling of nitro arenes with arylboronic acids.
  • Triethylamine-catalyzed three-component Hantzsch condensations.
  • Copper-catalyzed nitrations.
  • Oxidative mono-cleavage of dialkenes catalyzed by Trametes hirsuta.
  • Palladacycle-catalyzed cross-coupling of arylboronic acids with carboxylic anhydrides or acyl chlorides.
  • Palladium-catalyzed aerobic oxidative cross-coupling reactions.
  • The synthesis of sensitizers with dithiafulvenyl unit as electron donor for high-efficiency dye-sensitized solar cells.
  • The synthesis of a novel protein synthesis inhibitor active against Gram-positive bacteria.
  • The Suzuki aryl-aryl coupling of the upper rim of hexahomotrioxacalix[3]arene.
  • A rhodium-catalyzed cyclization, converting 1,5-enynes to cyclopentenes and spiro-cyclopentenes.

Other Notes

Contains varying amounts of anhydride

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shi-En Chen et al.
Analytica chimica acta, 1124, 40-51 (2020-06-15)
Glucosylsphingosine (GlcS) in plasma is considered to be a reliable biomarker of Gaucher disease. The detection difficulty of GlcS is that it is difficult to achieve simultaneous separation and quantification with its isomer galactosylsphingosine (GalS), a biomarker of Krabbe disease.
A new and simple synthetic approach to functionalized sulphone derivatives by the Suzuki-Miyaura cross-coupling reaction
Kotha S and Ghosh AK
Indian J. Chem. B, 45(1), 227-231 (2006)
Nora R Eibergen et al.
Chembiochem : a European journal of chemical biology, 13(4), 574-583 (2012-03-01)
In an effort to identify novel antibacterial chemotypes, we performed a whole-cell screen for inhibitors of Staphylococcus aureus growth and pursued those compounds with previously uncharacterized antibacterial activity. This process resulted in the identification of a benzothiazolium salt, ABTZ-1, that
Caroline E Paul et al.
Chemical communications (Cambridge, England), 48(27), 3303-3305 (2012-02-24)
The first report of a biocatalytic regioselective oxidative mono-cleavage of dialkenes was successfully achieved employing a cell-free enzyme preparation from Trametes hirsuta at the expense of molecular oxygen. Selected reactions were performed on a preparative scale affording high to excellent
Suzuki Coupling at the 2-Position of Densely Functionalized Pyrimidones
Colarusso, Stefania; et al.
Synthesis, 8, 1343-1350 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service