Skip to Content
Merck
All Photos(1)

Documents

E50205

Sigma-Aldrich

Ethyl 4,4,4-trifluoroacetoacetate

99%

Synonym(s):

ETFAA, Ethyl 3-oxo-4,4,4-trifluorobutyrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3COCH2CO2C2H5
CAS Number:
Molecular Weight:
184.11
Beilstein:
608353
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.375 (lit.)

bp

129-130 °C (lit.)

density

1.259 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)CC(=O)C(F)(F)F

InChI

1S/C6H7F3O3/c1-2-12-5(11)3-4(10)6(7,8)9/h2-3H2,1H3

InChI key

OCJKUQIPRNZDTK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ethyl 4,4,4-trifluoroacetoacetate (ETFAA) is a general reagent to synthesize enantiopure trifluoromethyl-functionalized products. Applications include:
  • Synthesis of (S)- and (R)-α-trifluoromethyl-aspartic acid and α- trifluoromethyl-serine from chiral CF3-oxazolidines, which is derived from ETFAA.
  • Enantiopure synthesis of trifluoromethyl-β-amino acid derivatives.
  • Synthesis of (2R)-2-trifluoromethyl-2-carboxyazetidine, (R)- and (S)-trifluoromethylhomoserines from oxazolidine intermediate obtained by condensing (R)-phenylglycinol with ETFAA.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

100.4 °F - closed cup

Flash Point(C)

38 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Concise synthesis of enantiopure (S)-and (R)-α-trifluoromethyl aspartic acid and α-trifluoromethyl serine from chiral trifluoromethyl oxazolidines (Fox) via a Strecker-type reaction.
Simon J, et al.
Tetrahedron Asymmetry, 22(3), 309-314 (2011)
Straightforward Synthesis of Novel Enantiopure α-Trifluoromethylated Azetidine 2-Carboxylic Acid and Homoserines.
Lensen N, et al.
Organic Letters, 17(2), 342-345 (2015)
Ethyl-4, 4, 4-trifluoroacetoacetate (ETFAA), a powerful building block for enantiopure chirons in trifluoromethyl-β-amino acid series.
Michaut V, et al.
Journal of Fluorine Chemistry, 128(8), 889-895 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service