249920
Silicon tetrachloride solution
1.0 M in methylene chloride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
SiCl4
CAS Number:
Molecular Weight:
169.90
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
vapor density
5.86 (vs air)
form
liquid
concentration
1.0 M in methylene chloride
density
1.34 g/mL at 25 °C
SMILES string
Cl[Si](Cl)(Cl)Cl
InChI
1S/Cl4Si/c1-5(2,3)4
InChI key
FDNAPBUWERUEDA-UHFFFAOYSA-N
Related Categories
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Inhalation - Carc. 2 - Eye Dam. 1 - Skin Corr. 1A - STOT SE 3
Target Organs
Central nervous system
Supplementary Hazards
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
D Khushalani et al.
Journal of nanoscience and nanotechnology, 1(2), 129-132 (2003-08-14)
Mesostructured silica-nanoparticle monolithic composites have been synthesized by dispersing prefabricated nanoparticles of gold or zeolite (silicalite) in ethanolic reaction mixtures containing SiCl4 and a Pluronic triblock copolymer template. Whereas silicalite nanoparticles were used directly, surface functionalization of the gold nanoparticles
Pulmonary interstitial emphysema: CT findings.
A C Kemper et al.
AJR. American journal of roentgenology, 172(6), 1642-1642 (1999-06-01)
Baker Jawabrah Al-Hourani et al.
Bioorganic & medicinal chemistry letters, 22(6), 2235-2238 (2012-02-22)
A series of novel 5-substituted 1H-tetrazoles as cyclooxygenase-2 (COX-2) inhibitors was prepared via treatment of various diaryl amides with tetrachlorosilane/sodium azide. All compounds were tested in cyclooxygenase (COX) assays in vitro to determine COX-1 and COX-2 inhibitory potency and selectivity.
Claudio Curti et al.
Organic letters, 13(17), 4738-4741 (2011-08-13)
Virtually perfect transmittal of the enolate reactivity up to five conjugated double bonds from the origin allows a series of furan-based silyloxypolyenes to add to aldehyde carbonyls at the most distant point of the molecule. Denmark's axially chiral bisphosphoramide/SiCl(4) combination
Sang Ho Lee et al.
Journal of the American Chemical Society, 127(25), 9071-9078 (2005-06-23)
A series of four asymmetrically aryl-substituted 9,9'-spiro-9-silabifluorene (SSF) derivatives, 2,2'-di-tert-butyl-7,7'-diphenyl-9,9'-spiro-9-silabifluorene (PhSSF), 2,2'-di-tert-butyl-7,7'-dipyridin-2-yl-9,9'-spiro-9-silabifluorene (PySSF), 2,2'-di-tert-butyl-7,7'-dibiphenyl-4-yl-9,9'-spiro-9-silabifluorene (BPhSSF), and 2,2'-di-tert-butyl-7,7'-bis(2',2' '-bipyridin-6-yl)-9,9'-spiro-9-silabifluorene (BPySSF) are prepared through the cyclization of the corresponding 2,2'-dilithiobiphenyls with silicon tetrachloride. These novel spiro-linked silacyclopentadienes (siloles) form transparent and stable
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service