Skip to Content
Merck
All Photos(1)

Key Documents

247480

Sigma-Aldrich

Domiphen bromide

97%

Synonym(s):

(Dodecyldimethyl-2-phenoxyethyl)ammonium bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)11N(CH3)2(CH2CH2OC6H5)Br
CAS Number:
Molecular Weight:
414.46
EC Number:
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

powder

mp

117-119 °C (lit.)

SMILES string

[Br-].CCCCCCCCCCCC[N+](C)(C)CCOc1ccccc1

InChI

1S/C22H40NO.BrH/c1-4-5-6-7-8-9-10-11-12-16-19-23(2,3)20-21-24-22-17-14-13-15-18-22;/h13-15,17-18H,4-12,16,19-21H2,1-3H3;1H/q+1;/p-1

InChI key

OJIYIVCMRYCWSE-UHFFFAOYSA-M

General description

Domiphen bromide (DMP) is a quaternary ammonium compound that belongs to the class of phenol ethers. It can be used as a cationic surfactant.

Application

  • Domiphen Bromide in Antimicrobial Applications: A clinical trial assessed the effectiveness of a mouthwash containing Domiphen Bromide among other quaternary ammonium compounds, showcasing its ability to significantly reduce gingivitis, supporting its use in dental health products (Rosa and Sturzenberger, 1976).
  • Toxicity and Safety Profile of Domiphen Bromide: A recent study explored the proarrhythmic effects induced by Domiphen Bromide in both in vitro and in vivo models, providing vital safety information crucial for its use in pharmaceutical applications (Long et al., 2021).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

F Scaglione et al.
International journal of clinical pharmacology research, 3(5), 357-361 (1983-01-01)
Thirty-one patients (21 M and 10 F), affected by acute infectious dental disease, were treated with domifen bromide (A) or placebo (Pl). It was found that A had a beneficial effect on the results of clinical controls and reduced the
[Antiseptic activity of domiphen bromide on the yeast Candida in the mouth].
P Goumas et al.
Stomatologia, 41(5), 249-257 (1984-09-01)
Separation of fulvic acid from soil extracts based on ion-pair formation with a cationic surfactant
Fukushima M, et al.
Analytical Sciences, 22(2), 229-233 (2006)
Thermophysical Studies on Molecular Interactions of Semicarbazide Hydrochloride/Domiphen Bromide in Aqueous Deep Eutectic Solvent Media at Various Temperatures
Majid Q, et al.
Journal of Chemical and Engineering Data, 67(10), 2974-2985 (2022)
Julien R G Navarro et al.
Nanotechnology, 23(46), 465602-465602 (2012-10-26)
A great number of works have focused their research on the synthesis, design and optical properties of gold nanoparticles for potential biological applications (bioimaging, biosensing). For this kind of application, sharp gold nanostructures appear to exhibit the more interesting features

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service