Skip to Content
Merck
All Photos(1)

Key Documents

D200603

Sigma-Aldrich

2,6-Dinitrotoluene

98%

Synonym(s):

1-Methyl-2,6-dinitrobenzene, 2,6-DNT, 2-Methyl-1,3-dinitrobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3(NO2)2
CAS Number:
Molecular Weight:
182.13
Beilstein:
2052046
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

crystals

mp

56-61 °C (lit.)

SMILES string

CC1=C([N+]([O-])=O)C=CC=C1[N+]([O-])=O

InChI

1S/C7H6N2O4/c1-5-6(8(10)11)3-2-4-7(5)9(12)13/h2-4H,1H3

InChI key

XTRDKALNCIHHNI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Carc. 1B - Muta. 2 - Repr. 2 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

404.1 °F - closed cup

Flash Point(C)

206.7 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kurt A Gust et al.
Toxicological sciences : an official journal of the Society of Toxicology, 110(1), 168-180 (2009-05-07)
Munitions constituents (MCs) including hexahydro-1,3,5-trinitro-1,3,5-triazine (RDX), 2,4,6-trinitrotoluene (TNT), and TNT derivatives are recognized to elicit aberrant neuromuscular responses in many species. The onset of seizures resulting in death was observed in the avian model Northern bobwhite after oral dosing with
Kevin A Thorn et al.
Environmental science & technology, 42(7), 2542-2550 (2008-05-29)
Large-scale aerobic windrow composting has been used to bioremediate washout lagoon soils contaminated with the explosives TNT (2,4,6-trinitrotoluene) and RDX (hexahydro-1,3,5-trinitro-1,3,5-triazine) at several sites within the United States. We previously used 15N NMR to investigate the reduction and binding of
Katerina M Dontsova et al.
Chemosphere, 77(4), 597-603 (2009-09-05)
Live-fire training exercises can result in particulate propellant contamination on military training ranges and can potentially contaminate ground water. This study was conducted to evaluate dissolution of the 2,4-dinitrotoluene (2,4-DNT) and 2,6-dinitrotoluene (2,6-DNT) from the propellant formulation, M1 (87.6% nitrocellulose
Hong Yang et al.
Chemosphere, 70(5), 791-799 (2007-09-04)
Dinitrotoluenes (DNTs) are widely used in the manufacturing of explosives and propellants hence causing contamination of several terrestrial and aquatic environments. The present study describes biotransformation of 2,4-DNT and 2,6-DNT in marine sediment sampled from a shipwreck site near Halifax
K M Dokken et al.
Journal of environmental quality, 40(3), 719-730 (2011-05-07)
Infrared microspectroscopy (IMS) is emerging as an important analytical tool for the structural analysis of biological tissue. This report describes the use of IMS coupled to a synchrotron source combined with principal components analysis (PCA) to monitor the fate and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service