234265
Sodium dihydrogencitrate
99%
Synonym(s):
Citric acid monosodium salt, Sodium citrate monobasic
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
HOC(COONa)(CH2COOH)2
CAS Number:
Molecular Weight:
214.11
Beilstein:
3920246
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
99%
form
solid
mp
212 °C (lit.)
solubility
water: soluble 53.5 g/L at 20 °C
functional group
carboxylic acid
hydroxyl
SMILES string
[Na+].OC(=O)CC(O)(CC([O-])=O)C(O)=O
InChI
1S/C6H8O7.Na/c7-3(8)1-6(13,5(11)12)2-4(9)10;/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);/q;+1/p-1
InChI key
HWPKGOGLCKPRLZ-UHFFFAOYSA-M
General description
Freezing temperature lowerings of aqueous solutions of sodium dihydrogencitrate has been investigated.
Application
Sodium dihydrogencitrate has been employed:
- as reductant and stabilizer during the synthesis of Fe3O4 Pt core/shell particles
- in characterization of carbon-based electrode materials as cathode in half-cell configuration
- in electrochemical reduction of proton at carbon fiber electrode in the presence and absence of Pt nanoparticles
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Electrochemistry of single nanoparticles via electrocatalytic amplification.
Bard AJ, et al.
Israel J. Chem., 50(3), 267-276 (3010)
Cryoscopic studies of aqueous solutions of tartaric acid, sodium hydrogen tartrate, potassium tartrate, sodium dihydrogen citrate, potassium dihydrogen citrate, disodium hydrogen citrate, sodium citrate and potassium citrate.
Apelblat A and Manzurola E.
The Journal of Chemical Thermodynamics, 35(8), 1225-1236 (2003)
The self-assembly, characterization and application of hemoglobin immobilized on Fe3O4@ Pt core-shell nanoparticles.
Fan H, et al.
Microchimica Acta, 168(3-4), 239-244 (2010)
Don-On Daniel Mak et al.
Cold Spring Harbor protocols, 2013(9), 880-884 (2013-09-05)
Nuclear patch-clamp experiments can be performed with intact nuclei or with nuclei from which the outer nuclear membrane has been removed. This protocol presents procedures for harvesting different types of cultured cells, isolating nuclei, and exposing the inner nuclear membrane
Stefanie Rubenwolf et al.
Biosensors & bioelectronics, 26(2), 841-845 (2010-07-16)
Direct electron transfer from carbon electrodes to adsorbed laccase (EC 1.10.3.2) from Trametes versicolor is widely used to enable mediatorless enzymatic biofuel cell cathodes. However, data published so far are poorly comparable in terms of oxygen reduction performance. We thus
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service