Skip to Content
Merck
All Photos(3)

Key Documents

S6776

Sigma-Aldrich

Sodium fluoride

BioReagent, suitable for insect cell culture, ≥99%

Synonym(s):

NaF

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaF
CAS Number:
Molecular Weight:
41.99
EC Number:
MDL number:
UNSPSC Code:
12352302
eCl@ss:
38050104
PubChem Substance ID:
NACRES:
NA.71

vapor pressure

1.4 mmHg ( 0 °C)

product line

BioReagent

Assay

≥99%

technique(s)

cell culture | insect: suitable

mp

993 °C (lit.)

SMILES string

[F-].[Na+]

InChI

1S/FH.Na/h1H;/q;+1/p-1

InChI key

PUZPDOWCWNUUKD-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2

Supplementary Hazards

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Naoomi Tominaga et al.
Nature communications, 6, 6716-6716 (2015-04-02)
Brain metastasis is an important cause of mortality in breast cancer patients. A key event during brain metastasis is the migration of cancer cells through blood-brain barrier (BBB). However, the molecular mechanism behind the passage through this natural barrier remains
Ryan D Espy et al.
Analytical chemistry, 86(15), 7712-7718 (2014-06-28)
Determination of eight drugs of abuse in blood has been performed using paper spray or extraction spray mass spectrometry in under 2 min with minimal sample preparation. A method has been optimized for quantification of amphetamine, methamphetamine, 3,4-methylenedioxyamphetamine (MDA), 3,4-methylenedioxy-N-methylamphetamine
E E Vincent et al.
Oncogene, 34(28), 3627-3639 (2014-09-23)
As a sensor of cellular energy status, the AMP-activated protein kinase (AMPK) is believed to act in opposition to the metabolic phenotypes favored by proliferating tumor cells. Consequently, compounds known to activate AMPK have been proposed as cancer therapeutics. However
Hannah Nguyen et al.
The Journal of biological chemistry, 290(22), 13641-13653 (2015-04-01)
SMYD2 is a lysine methyltransferase that catalyzes the monomethylation of several protein substrates including p53. SMYD2 is overexpressed in a significant percentage of esophageal squamous primary carcinomas, and that overexpression correlates with poor patient survival. However, the mechanism(s) by which
Pierre-Olivier Strale et al.
The Journal of cell biology, 210(2), 333-346 (2015-07-22)
Oligomerization of cadherins could provide the stability to ensure tissue cohesion. Cadherins mediate cell-cell adhesion by forming trans-interactions. They form cis-interactions whose role could be essential to stabilize intercellular junctions by shifting cadherin clusters from a fluid to an ordered

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service