Skip to Content
Merck
All Photos(2)

Key Documents

229962

Sigma-Aldrich

Thallium(I) chloride

99.999% trace metals basis

Synonym(s):

Thallous chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
TlCl
CAS Number:
Molecular Weight:
239.84
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

10 mmHg ( 517 °C)

Quality Level

Assay

99.999% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: thallium

impurities

≤15.0 ppm Trace Metal Analysis

bp

720 °C/1 atm (lit.)

mp

430 °C (lit.)

solubility

ethanol: insoluble(lit.)

density

7 g/mL at 25 °C (lit.)

SMILES string

Cl[Tl]

InChI

1S/ClH.Tl/h1H;/q;+1/p-1

InChI key

GBECUEIQVRDUKB-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Used to prepare cage, dinuclear complexes (TI-cyclodiphosphazanes) which possess interesting metal-metal interactions.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Chronic 2 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zekiye Hasbek et al.
Nuclear medicine communications, 30(10), 779-788 (2009-08-06)
The aim of this study was to evaluate the myocardial viability in nondiabetic patients with chronic coronary artery disease (CCAD) or past myocardial infarction (MI), using thallium-201 infusion myocardial perfusion single-photon emission computed tomography (MPSPECT) imaging after oral glucose application
Barbra E Backus et al.
Journal of nuclear cardiology : official publication of the American Society of Nuclear Cardiology, 16(1), 97-104 (2009-01-20)
To define the physical and clinical reproducibility of (201)Tl myocardial perfusion SPECT (MPS), this study assesses the variation between two repeated rest (201)Tl MPS with repositioning only, with a two-hour time interval and with phantom measurements as a reference. Three
P A Howard
DICP : the annals of pharmacotherapy, 25(10), 1085-1091 (1991-10-01)
An intravenous form of dipyridamole has been approved for cardiac stress testing with thallous chloride Tl 201 (201Tl) imaging. The procedure provides an alternative method for stress testing of patients who cannot perform strenuous physical exercise because of physical limitations
Kimie Asao et al.
Annals of nuclear medicine, 26(2), 123-130 (2011-10-29)
The interpolated projection data estimation processing (IPDE) method increases the amount of projection data by interpolation of the projection data. We examined the usefulness of the IPDE method for (201)Tl myocardial perfusion imaging (MPI) single photon emission computed tomography (SPECT)
Biological safety of nasal thallium-201 administration: a preclinical study for olfacto-scintigraphy.
Kohshin Washiyama et al.
Journal of radiation research, 52(4), 450-455 (2011-09-10)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service